Skip to Content

AG549

α-synuclein 61-140, human

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
12352203
NACRES:
NA.77
eCl@ss:
32160702
Form:
powder
Assay:
>95% (SDS-PAGE)
Biological source:
human
Mol wt:
Mw 8,460 Da
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

human

Quality Segment

assay

>95% (SDS-PAGE)

form

powder

mol wt

Mw 8,460 Da

manufacturer/tradename

Chemicon®

technique(s)

cell based assay: suitable

NCBI accession no.

UniProt accession no.

shipped in

dry ice

Gene Information

human ... SNCA(6622)

General description

Alpha-synuclein is encoded by the SNCA gene (also known as NACP, PARK1) in human. There are two other known synuclein family members, beta- and gamma-synuclein. Interest in synuclein research started with the discovery of alpha-synuclein mutation in several families with autosomal dominant Parkinson′s disease (PD). Alpha-synuclein is abundantly expressed in the brain and is found in a classic amyloid fibril form within the intra-neuronal Lewy body deposits of PD. The physiological function of synucleins is not well understood, but appears to involve membrane interactions, and in particular reversible binding to synaptic vesicle membranes. Contradictory evidences regarding inhibitory action of alpha-synuclein against phospholipase D (PLD) have been reported (PMID 11821392 & 19146388). Clone Syn211, Cat. No. 36-008-C detects epitope in the region from amino acid 121 to 125 and has been used together with clone LB509, Cat. No. MABN824, to characterize the digestion pattern of different forms of in vitro prepared alpha-synclulein aggregates (Guo, J.L., et al. 2013; PMID 23827677).
MEQVTNVGGAVVTGVTAVAQKTVEGAGSIAAATGFVKKDQLGKNEE-
GAPQEGILEDMPVDPDNEAYEMPSEEGYQDYEPEA
An additional amino acid (Met) is attached at the N-terminus.
Product Source: E. coli

Application

Research Category
Neuroscience
Research Sub Category
Neurodegenerative Diseases

Physical form

White lyophilized powder. Resuspend in sterile water at concentration of 1 mg/mL. This will give you a final of 20 mM Tris/HCI, pH 7.5, 100 mM NaCl.

Preparation Note

Maintain lyophilized material at -20°C for up to 12 months after date of receipt. After reconstitution maintain at -20°C to -70°C for up to 2 weeks in undiluted aliquots. Avoid freeze/thaw cycles to avoid aggregation.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.


Storage Class

11 - Combustible Solids

wgk

WGK 1



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library