Skip to Content
Merck
All Photos(1)

Key Documents

ABT171

Sigma-Aldrich

Anti-alpha Tubulin Antibody, tyrosinated

serum, from rabbit

Synonym(s):

Tubulin alpha-1A chain, Alpha-tubulin 3, Tubulin B-alpha-1, Tubulin alpha-3 chain

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

serum

antibody product type

primary antibodies

clone

polyclonal

species reactivity

monkey, mouse

species reactivity (predicted by homology)

rat (based on 100% sequence homology), human (based on 100% sequence homology)

technique(s)

ELISA: suitable
immunocytochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... TUBA1A(7846)

General description

Microtubules are involved in a wide variety of cellular activities ranging from mitosis and transport events to cell movement and the maintenance of cell shape. Tubulin itself is a globular protein which consists of two polypeptides (alpha and beta tubulin). Alpha and beta tubulin dimers are assembled to 13 protofilaments that form a microtubule of 22 nm diameter. Tyrosine ligase adds a C-terminal tyrosine to monomeric alpha tubulin. Assembled microtubules can be detyrosinated by a cytoskeleton associated carboxypeptidase. Detyrosinated alpha tubulin is referred to as Glu-tubulin. Another post-translational modification of detyrosinated alpha tubulin is C-terminal polyglutamylation which is characteristic for microtubules in neuronal cells and the mitotic spindle.

Specificity

This antibody detects the tyrosinated form of alpha Tubulin.

Immunogen

KLH-conjugated linear peptide corresponding to human tyrosinated alpha Tubulin.

Application

Detect Tubulin using this rabbit polyclonal antibody, Anti-alpha Tubulin Antibody, tyrosinated validated for use in western blotting, ELISA & ICC.
Western Blotting Analysis: A representative lot from an independent laboratory specifically detected tyrosinated alpha Tubulin and not nontyrosinated alpha Tubulin using purified porcine tubulin (Gundersen, G. G., et al. (1984). Cell. 38(3):779-89.).

ELISA Analysis: A representative lot from an independent laboratory specifically detected tyrosinated alpha Tubulin, in an indirect ELISA and in a competitive ELISA (Gundersen, G. G., et al. (1984). Cell. 38(3):779-89.).

Immunocytochemistry Analysis: A representative lot from an independent laboratory specifically detected tyrosinated alpha Tubulin and not nontyrosinated alpha Tubulin in TC-7 cells in interphase (Gundersen, G. G., et al. (1984). Cell. 38(3):779-89.).

Quality

Evaluated by Western Blotting in NIH/3T3 cell lysate, which were untreated or treated with Pancreatic Carboxypeptidase (PCA).

Western Blotting Analysis: A 1:2,000 dilution of this antibody detected alpha Tubulin, tyrosinated in 10 µg of untreated NIH/3T3 cell lysate and demonstrated a loss of signal in PCA treated NIH/3T3 cell lysate.

Target description

~51 kDa observed. Uncharacterized band(s) may be observed in some cell lysates.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Siwen Wu et al.
Endocrinology, 162(11) (2021-08-04)
Studies suggest that HIV-1 invades the testis through initial permeation of the blood-testis barrier (BTB). The selectivity of the BTB to antiretroviral drugs makes this site a sanctuary for the virus. Little is known about how HIV-1 crosses the BTB
Signe S Tofteng et al.
Acta physiologica (Oxford, England), 235(4), e13828-e13828 (2022-05-12)
Cyclooxygenase-2 (COX-2) activity protects against oxidative stress and apoptosis early in experimental kidney injury. The present study was designed to test the hypothesis that COX-2 activity attenuates fibrosis and preserves microvasculature in injured kidney. The murine unilateral ureteral-obstruction (UUO) model
Andrew W Mathewson et al.
Developmental biology, 452(1), 21-33 (2019-04-29)
The asymmetric localization of planar cell polarity (PCP) proteins is essential for the establishment of many planar polarized cellular processes, but the mechanisms that maintain these asymmetric distributions remain poorly understood. A body of evidence has tied oriented subapical microtubules
Pradeepa Jayachandran et al.
Neural development, 11, 1-1 (2016-01-20)
Shaping of the neural tube, the precursor of the brain and spinal cord, involves narrowing and elongation of the neural tissue, concomitantly with other morphogenetic changes that contribue to this process. In zebrafish, medial displacement of neural cells (neural convergence
Nan Tian et al.
Journal of neurochemistry, 152(5), 542-555 (2019-11-11)
Axon degeneration and axonal loss is a feature of neurodegenerative disease and injury and occurs via programmed pathways that are distinct from cell death pathways. While the pathways of axonal loss following axon severing are well described, less is known

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service