Skip to Content
Merck
All Photos(2)

Key Documents

ABT112

Sigma-Aldrich

Anti-Lysyl Oxidase (LOX) Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

Protein-lysine 6-oxidase, Lysyl oxidase

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

mouse, human

species reactivity (predicted by homology)

rat (based on 100% sequence homology)

technique(s)

immunohistochemistry: suitable (paraffin)
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... LOX(4015)

General description

Lysyl Oxidase (Protein-lysine 6-oxidase; LOX) is a member of the lysyl oxidase family and a product of the LOX gene. Lysyl oxidase is an extracellular-matrix oxidative enzyme characterized by its C-terminal LOX catalytic domain. In the presence of copper, lysyl oxidase removes amine groups from lysine residues of target substrates which include precursors of collagen and elastin. This deamination reaction produces reactive lysine residues which can then interact with similarly activated species to form covalent cross-links. Lysyl oxidase is found predominantly in the kidney, pancreas, heart, skeletal muscle, and placenta. Defective lysyl oxidase proteins play a role in cutis laxa autosomal recessive type 1 condition. Other studies have also suggested that lysyl oxidase may also be involved in intracellular signaling and may function as a tumor suppressor.

Immunogen

KLH-conjugated linear peptide corresponding to Lysyl Oxidase (LOX).

Application

Anti-Lysyl Oxidase (LOX) Antibody is an antibody against Lysyl Oxidase (LOX) for use in Western Blotting, IHC(P).
Immunohistochemistry Analysis: A 1:100 dilution from a representative lot detected Lysyl Oxidase (LOX) in adenocarcinoma cells of human prostate tissues.

Quality

Evaluated by Western Blot in NIH/3T3 nuclear extract.

Western Blot Analysis: A 1:1,000 dilution of this antibody detected Lysyl Oxidase (LOX) in 10 µg of NIH/3T3 nuclear extract.

Target description

~46 kDa observed

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Heyong Yin et al.
Biomedical materials (Bristol, England), 13(3), 034107-034107 (2018-02-09)
Thermosensitive hydrogels have been studied for potential application as promising alternative cell carriers in cell-based regenerative therapies. In this study, a thermosensitive butane diisocyanate (BDI)-collagen hydrogel (BC hydrogel) was designed as an injectable cell delivery carrier of tendon stem/progenitor cells
Targeting the LOX/hypoxia axis reverses many of the features that make pancreatic cancer deadly: inhibition of LOX abrogates metastasis and enhances drug efficacy.
Miller, BW; Morton, JP; Pinese, M; Saturno, G; Jamieson, NB; McGhee, E; Timpson, P; Leach et al.
EMBO Molecular Medicine null
Masaru Kaku et al.
Journal of cellular physiology, 231(4), 926-933 (2015-09-19)
Type I collagen, a major extracellular component of the periodontal ligament (PDL), is post-translationally modified by a series of specific enzymes. Among the collagen-modifying enzymes, lysyl oxidase (LOX) is essential to initiate collagen cross-linking and lysyl hydroxylases (LHs) to regulate
Rolf Schreckenberg et al.
Frontiers in physiology, 8, 556-556 (2017-08-22)
Purpose: According to the current therapeutic guidelines of the WHO physical activity and exercise are recommended as first-line therapy of arterial hypertension. Previous results lead to the conclusion, however, that hearts of spontaneously hypertensive rats (SHR) with established hypertension cannot
Rohit Shetty et al.
Molecular vision, 21, 12-25 (2015-01-17)
Keratoconus (KC) is characterized by progressive vision loss due to corneal thinning and structural abnormalities. It is hypothesized that KC is caused by deregulated collagen levels and collagen fibril-maturating enzyme lysyl oxidase (LOX). Further, it is currently not understood whether

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service