Skip to Content
Merck

ABS818

Anti-Ovalbumin (Egg White)

from rabbit, purified by affinity chromatography

Synonym(s):

Allergen Gal d II, Egg albumin, Plakalbumin

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
eCl@ss:
32160702
Conjugate:
unconjugated
Clone:
polyclonal
Application:
WB
Citations:
1
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

rabbit

Quality Segment

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

chicken

packaging

antibody small pack of 25 μL

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

ambient

target post-translational modification

unmodified

Gene Information

chicken ... Oval(396058)

General description

Ovalbumin (UniProt: P01012; also known as Allergen Gal d II, Egg albumin, Plakalbumin, Gal d 2) is encoded by the SERPINB14 gene (Gene ID: 396058) in chicken. Ovalbumin is a member of the serine protein inhibitor (serpin) super family, but it considered as a non-inhibitory serpin. It is a storage protein of egg white and may make up about 60% of the total protein in egg white. Ovalbumin provides a major interest for study because its structure provides a model for the structure of the inhibitory serpins. It is known to chelate heavy metals and traps the metal ions within the sulfhydryl bonds of the protein, which prevents the absorption of the metals into the gastrointestinal tract and prevents poisoning.
~43 kDa observed; 42.88 kDa calculated. Uncharacterized bands may be observed in some lysate(s).

Immunogen

Full-length purified chicken egg ovalbumin.

Application

Anti-Ovalbumin (Egg White) Antibody, Cat. No. ABS818, is a rabbit polyclonal antibody that detects chicken egg ovalbumin and has been tested for use in Western Blotting.

Biochem/physiol Actions

This rabbit polyclonal antibody detects chicken egg ovalbumin.

Analysis Note

Evaluated by Western Blotting in chicken egg ovalbumin.

Western Blotting Analysis: A 1:10,000 dilution of this antibody detected Ovalbumin in chicken eggs.

Other Notes

Concentration: Please refer to lot specific datasheet.


Still not finding the right product?

Try our Product Selector Tool to narrow your options


Storage Class

12 - Non Combustible Liquids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library