Skip to Content
Merck
All Photos(5)

Key Documents

ABN99

Sigma-Aldrich

Anti-phospho NR1 (Ser897) Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

glutamate receptor, ionotropic, N-methyl D-aspartate 1, N-methyl-D-aspartate receptor subunit NR1, glutamate [NMDA] receptor subunit zeta 1, N-methyl-D-aspartate receptor channel, subunit zeta-1, glutamate [NMDA] receptor subunit zeta-1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41
clone:
polyclonal
application:
IHC
IP
WB
species reactivity:
rat
technique(s):
immunohistochemistry: suitable
immunoprecipitation (IP): suitable
western blot: suitable
citations:
18

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

rat

species reactivity (predicted by homology)

mouse (based on 100% sequence homology), human (based on 100% sequence homology)

technique(s)

immunohistochemistry: suitable
immunoprecipitation (IP): suitable
western blot: suitable

UniProt accession no.

shipped in

wet ice

target post-translational modification

phosphorylation (pSer897)

Gene Information

human ... GRIN1(2902)
mouse ... Grin1(14810)
rat ... Grin1(24408)

General description

NR1 (NMDA1or NMDAR1, also known as GRIN1) along with the NR2 subunit forms a heterodimer that make up the NMDA receptor. The NMDA receptor (NMDAR) is characterized as a ionotropic receptor for glutamate. As the name suggests, (NMDA (N-methyl D-aspartate) is an agonist for the receptor. A distinct characteristic of the NMDA receptor is that it is both ligand-gated and voltage-dependent based on its sensitivity to magnesium. Activated NMDA receptors result in the opening of an ion channel that is nonselective to cations. The result is the flow of Na+ and small amounts of Ca2+ ions into the cell and K+ out of the cell. Calcium flux by the NMDARs has been implicated in synaptic plasticity, a mechanism in the cell for learning and memory.

Specificity

This antibody recognizes NR1 when phosphorylated at Ser897.

Immunogen

Epitope: Phosphorylated Ser897
KLH-conjugated linear peptide corresponding to rat NR1 when phosphorylated at Ser897.

Application

Immunohistochemistry Analysis: 1:50 dilution from a previous lot detected NR1 in normal rat hippocampus.
Immunoprecipitation Analysis: 10 µg from a previous lot immunoprecipitated NR1 from rat brain microsomal RIPA lysate.
Western Blot Analysis: 2 µg/mL from a previous lot detected NR1 in Lambda phosphatase untreated and treated rat brain microsomal tissue lysate.
Research Category
Neuroscience
Research Sub Category
Signaling Neuroscience
This Anti-phospho NR1 (Ser897) Antibody is validated for use in WB, IP, IH for the detection of phospho NR1 (Ser897).

Quality

Evaluated by Western Blot in rat brain microsomal tissue lysate.
Western Blot Analysis: 1 µg/mL of this antibody detected NR1 on 10 µg of rat brain microsomal tissue lysate.

Target description

~ 120 kDa

Linkage

Replaces: 06-641

Physical form

Affinity purified
Purified rabbit polyclonal in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
Rat brain microsomal tissue lysate

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Regular physical activity prevents development of chronic pain and activation of central neurons.
Sluka, KA; O'Donnell, JM; Danielson, J; Rasmussen, LA
Journal of Applied Physiology (1985)
Lucas V Lima et al.
Pain, 158(9), 1697-1710 (2017-06-18)
Regular physical activity prevents the development of chronic muscle pain through the modulation of central mechanisms that involve rostral ventromedial medulla (RVM). We tested if pharmacological blockade or genetic deletion of mu-opioid receptors in physically active mice modulates excitatory and
Wenjuan Ru et al.
Journal of neuroimmune pharmacology : the official journal of the Society on NeuroImmune Pharmacology, 11(1), 182-191 (2015-11-20)
HIV-1 envelope glycoprotein gp120 (gp120) is a major virulence protein implicated in the pathogenesis of HIV-associated neurocognitive disorders (HAND). Although gp120 has been suggested to cause synaptic and neuronal injuries by disrupting NMDA receptor (NMDAR) function, the underlying mechanism is
Jinseok Yeo et al.
Pain research & management, 2017, 7394626-7394626 (2017-12-07)
The chronic postischemia pain (CPIP) model is an animal model using ischemia/reperfusion injury that mimics the symptoms of complex regional pain syndrome type I. Glutathione (GSH) prevents ischemia/reperfusion injury by scavenging free radicals. We conducted this study to investigate the
Jinseok Yeo et al.
Journal of pain research, 11, 3025-3030 (2018-12-21)
Complex regional pain syndrome type 1 (CRPS I) is an intractable neuropathic pain syndrome. Chronic post-ischemia pain (CPIP) model is an animal model of CRPS I which is produced by ischemia-reperfusion (IR) injury of the hind limb. Dexmedetomidine (DEX) is

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service