Skip to Content
Merck
All Photos(2)

Documents

ABN904

Sigma-Aldrich

Anti-GAD 65/67

from rabbit, purified by affinity chromatography

Synonym(s):

Glutamate decarboxylase 2/Glutamate decarboxylase 1, 65 kDa/67kDa glutamic acid decarboxylase, GAD-65/67, Glutamate decarboxylase 65/67 kDa isoform

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

rat, mouse, human

packaging

antibody small pack of 25 μg

technique(s)

immunohistochemistry: suitable (paraffin)
western blot: suitable

isotype

IgG

NCBI accession no.

UniProt accession no.

shipped in

ambient

target post-translational modification

unmodified

Gene Information

General description

Glutamate decarboxylase 2/Glutamate decarboxylase 1 (UniProt: Q05329/Q99259; also known as 65 kDa/67kDa glutamic acid decarboxylase, GAD-65/67, Glutamate decarboxylase 65/67 kDa isoform) is encoded by GAD2/GAD1 (also known as GAD65/GAD67) gene (Gene ID: 2572/2571) in human. Glutamate decarboxylase isoforms are expressed in the brain where it is involved in gamma-aminobutyric acid (GABA) synthesis. Glutamate decarboxylase 2 is also expressed in pancreatic tissue. Both GAD65 and GAD67 synthesize GABA at different locations in the cell and at different developmental stages. Usually, GAD1 is distributed evenly throughout the cell while isoform GAD2 is mainly localized to nerve terminals. GAD1 synthesizes GABA for neuronal activity that is related to as synaptogenesis and protection from neural injury. However, GAD2 isoform synthesizes GABA for neurotransmission activity.

Specificity

This rabbit polyclonal antibody detects GAD2 and GAD1 in human, Mouse, and Rat. It targets an epitope within 14 amino acids from the C-terminal region.

Immunogen

Epitope: C-terminus
KLH-conjugated linear peptide corresponding to 14 amino acids from the C-terminal region of human glutamate decarboxylase 2 (GAD2).

Application

Anti-GAD 65/67 Antibody, Cat. No. ABN904, is a highly specific rabbit polyclonal antibody that targets Human, Mouse, and Rat GAD2 and GAD1 and has been tested in Immunohistochemistry (Paraffin) and Western Blotting.
Immunohistochemistry Analysis: A 1:1,000 dilution from a representative lot detected GAD 65/67 in human cerebellum, mouse cerebral cortex, and rat cerebral cortex tissues.
Research Category
Neuroscience

Quality

Evaluated by Western Blotting in human brain tissue lysate.

Western Blotting Analysis: 2 µg/mL of this antibody detected GAD 65/67 in 10 µg of human brain tissue lysate.

Target description

~60/62kDa observed; 65.41/66.90 kDa calculated, respectively for GAD2 and GAD1. Uncharacterized bands may be observed in some lysate(s).

Physical form

Affinity Purified
Purified rabbit polyclonal antibody in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

does not flash

Flash Point(C)

does not flash


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Khaggeswar Bheemanapally et al.
Scientific reports, 11(1), 16079-16079 (2021-08-11)
Astrocyte glycogen, the primary energy reserve in brain, undergoes continuous remodeling by glucose passage through the glycogen shunt prior to conversion to the oxidizable energy fuel L-lactate. Glucogenic amino acids (GAAs) are a potential non-glucose energy source during neuro-metabolic instability.
Ayed A Alshamrani et al.
Neuropeptides, 82, 102055-102055 (2020-05-27)
The brain glycogen reserve is a source of oxidizable substrate fuel. Lactoprivic-sensitive hindbrain A2 noradrenergic neurons provide crucial metabolic-sensory input to downstream hypothalamic glucose-regulatory structures. Current research examined whether hindbrain glycogen fuel supply impacts A2 energy stability and governance of
Md Main Uddin et al.
BMC neuroscience, 21(1), 51-51 (2020-11-27)
Ventromedial hypothalamic nucleus (VMN) gluco-regulatory transmission is subject to sex-specific control by estradiol. The VMN is characterized by high levels of aromatase expression. The aromatase inhibitor letrozole (LZ) was used with high-resolution microdissection/Western blot techniques to address the hypothesis that
Abdulrahman Alhamyani et al.
AIMS neuroscience, 8(4), 510-525 (2021-12-09)
Gamma-aminobutyric acid (GABA) acts on ventromedial hypothalamic targets to suppress counter-regulatory hormone release, thereby lowering blood glucose. Maladaptive up-regulation of GABA signaling is implicated in impaired counter-regulatory outflow during recurring insulin-induced hypoglycemia (RIIH). Ventromedial hypothalamic nucleus (VMN) GABAergic neurons express
Karen P Briski et al.
Neuropeptides, 77, 101962-101962 (2019-09-07)
Hindbrain energy state shapes hypothalamic control of glucostasis. Dorsal vagal complex (DVC) L-lactate deficiency is a potent glucose-stimulatory signal that triggers neuronal transcriptional activation in key hypothalamic metabolic loci. The energy gauge AMPK is activated in DVC metabolic-sensory A2 noradrenergic

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service