Skip to Content
Merck
All Photos(1)

Documents

ABN78C3

Sigma-Aldrich

Anti-NeuN (rabbit) Antibody, Cy3 Conjugate

from rabbit, CY3 conjugate

Synonym(s):

NEUronal Nuclei, Neuronal Nuclei

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

CY3 conjugate

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

species reactivity

rat, mouse

technique(s)

immunocytochemistry: suitable

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

General description

ABN78 is a rabbit polyclonal version of the Anti-NeuN, clone A60 (MAB377), NeuN is a highly characterized and cited mouse monoclonal antibody that specifically recognizes the DNA-binding, neuron-specific protein NeuN, which is present in most CNS and PNS neuronal cell types of all vertebrates tested. NeuN protein distributions are apparently restricted to neuronal nuclei, perikarya and some proximal neuronal processes in both fetal and adult brain although, some neurons fail to be recognized by NeuN at all ages: INL retinal cells, Cajal-Retzius cells, Purkinje cells, inferior olivary and dentate nucleus neurons, and sympathetic ganglion cells are examples. Immunohistochemically detectable NeuN protein first appears at developmental timepoints that correspond with the withdrawal of the neuron from the cell cycle and/or with the initiation of terminal differentiation of the neuron. Immunoreactivity appears around E9.5 in the mouse neural tube and is extensive throughout the developing nervous system by E12.5. Strong nuclear staining suggests a nuclear regulatory protein function; however, no evidence currently exists as to whether the NeuN protein antigen has a function in the distal cytoplasm or whether it is merely synthesized there before being transported back into the nucleus. No difference between protein isolated from purified nuclei and whole brain extract on immunoblots has been found.

Specificity

The unconjugated rabbit Anti-NeuN (Catalog No. ABN78) has been shown to work in human.
This antibody recognizes NeuN at the N-terminus.

Immunogen

Epitope: N-terminus
GST-tagged recombinant protein corresponding to mouse NeuN.

Application

Anti-NeuN (rabbit) Antibody, Cy3 Conjugate is an antibody against NeuN (rabbit) for use in IC.
Research Category
Neuroscience
Research Sub Category
Developmental Neuroscience

Quality

Evaluated by Immunocytochemistry in rat E18 cortex cells.

Immunocytochemistry Analysis: A 1:100 dilution of this antibody detected NeuN in rat E18 cortex cells.

Target description

The unconjugated parent antibody (Cat. No. ABN78) has an observed molecular weight at ~46/48 kDa An uncharacterized band appears at ~62 kDa in some lysates.

Physical form

Affinity purified
Purified rabbit polyclonal conjugated to Cy3 in PBS with 0.1% sodium azide and 15 mg/mL BSA.

Storage and Stability

Maintain refrigerated at 2-8 °C protected from light in undiluted aliquots for up to 6 months from date of receipt.

Analysis Note

Control
Rat E18 cortex cells

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Ayanda Ngwenya et al.
Brain, behavior and evolution, 87(1), 19-38 (2016-02-26)
It is generally believed that animals with larger bodies require larger brains, composed of more neurons. Across mammalian species, there is a correlation between body mass and the number of brain neurons, albeit with low allometric exponents. If larger bodies
Débora Jardim-Messeder et al.
Frontiers in neuroanatomy, 11, 118-118 (2018-01-10)
Carnivorans are a diverse group of mammals that includes carnivorous, omnivorous and herbivorous, domesticated and wild species, with a large range of brain sizes. Carnivory is one of several factors expected to be cognitively demanding for carnivorans due to a
Joanna Jacob et al.
The Journal of comparative neurology, 529(14), 3375-3388 (2021-06-03)
With rates of psychiatric illnesses such as depression continuing to rise, additional preclinical models are needed to facilitate translational neuroscience research. In the current study, the raccoon (Procyon lotor) was investigated due to its similarities with primate brains, including comparable
Kunzhang Lin et al.
Molecular brain, 13(1), 45-45 (2020-03-22)
Understanding the connecting structure of brain network is the basis to reveal the principle of the brain function and elucidate the mechanism of brain diseases. Trans-synaptic tracing with neurotropic viruses has become one of the most effective technologies to dissect
Sandra E Dos Santos et al.
Brain, behavior and evolution, 89(1), 48-63 (2017-01-27)
In the effort to understand the evolution of mammalian brains, we have found that common relationships between brain structure mass and numbers of nonneuronal (glial and vascular) cells apply across eutherian mammals, but brain structure mass scales differently with numbers

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service