Skip to Content
Merck
All Photos(2)

Key Documents

ABN78A4

Sigma-Aldrich

Anti-NeuN (rabbit) Antibody, Alexa Fluor 488 Conjugate

from rabbit, ALEXA FLUOR 488

Synonym(s):

NEUronal Nuclei, Neuronal Nuclei

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

ALEXA FLUOR 488

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

species reactivity

rat, mouse

technique(s)

immunocytochemistry: suitable
immunohistochemistry: suitable

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

General description

ABN78 is a rabbit polyclonal version of the Anti-NeuN, clone A60 (MAB377), NeuN is a highly characterized and cited mouse monoclonal antibody that specifically recognizes the DNA-binding, neuron-specific protein NeuN, which is present in most CNS and PNS neuronal cell types of all vertebrates tested. NeuN protein distributions are apparently restricted to neuronal nuclei, perikarya and some proximal neuronal processes in both fetal and adult brain although, some neurons fail to be recognized by NeuN at all ages: INL retinal cells, Cajal-Retzius cells, Purkinje cells, inferior olivary and dentate nucleus neurons, and sympathetic ganglion cells are examples. Immunohistochemically detectable NeuN protein first appears at developmental timepoints that correspond with the withdrawal of the neuron from the cell cycle and/or with the initiation of terminal differentiation of the neuron. Immunoreactivity appears around E9.5 in the mouse neural tube and is extensive throughout the developing nervous system by E12.5. Strong nuclear staining suggests a nuclear regulatory protein function; however, no evidence currently exists as to whether the NeuN protein antigen has a function in the distal cytoplasm or whether it is merely synthesized there before being transported back into the nucleus. No difference between protein isolated from purified nuclei and whole brain extract on immunoblots has been found.

Specificity

The unconjugated rabbit Anti-NeuN (Catalog No. ABN78) has been shown to work in human.
This antibody recognizes NeuN at the N-terminus.

Immunogen

Epitope: N-terminus
GST-tagged recombinant protein corresponding to mouse NeuN.

Application

Detect NeuN (rabbit) using this Anti-NeuN (rabbit) Antibody, Alexa Fluor 488 Conjugate validated for use in IC, IH.
Immunocytochemistry analysis: A 1:100 dilution from a representative lot detected NeuN in rat E18 primary cortex cells.
Research Category
Neuroscience
Research Sub Category
Developmental Neuroscience

Quality

Evaluated by Immunohistochemistry in adult mouse brain tissue.

Immunohistochemistry Analysis: A 1:100 dilution of this antibody detected NeuN in adult mouse brain tissue.

Target description

The unconjugated parent antibody (Cat. No. ABN78) has an observed molecular weight at ~46/48 kDa An uncharacterized band appears at ~62 kDa in some lysates.

Physical form

Affinity purified
Purified rabbit polyclonal conjugated to Alexa Fluor 488 in PBS with 0.1% sodium azide and 15 mg/mL BSA.

Storage and Stability

Maintain refrigerated at 2-8 °C protected from light in undiluted aliquots for up to 6 months from date of receipt.

Analysis Note

Control
Adult mouse brain tissue

Legal Information

ALEXA FLUOR is a trademark of Life Technologies

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Estrogen receptor alpha deficiency protects against development of cognitive impairment in murine lupus.
Cunningham, MA; Wirth, JR; Freeman, LR; Boger, HA; Granholm, AC; Gilkeson, GS
Journal of Neuroinflammation null
Amanda C Mitchell et al.
Nature communications, 7, 12743-12743 (2016-09-07)
Neuronal epigenomes, including chromosomal loopings moving distal cis-regulatory elements into proximity of target genes, could serve as molecular proxy linking present-day-behaviour to past exposures. However, longitudinal assessment of chromatin state is challenging, because conventional chromosome conformation capture assays essentially provide
Mingfei Zhao et al.
Molecular medicine reports, 13(1), 13-20 (2015-11-18)
Traumatic brain injury (TBI) leads to permanent neurological impairment, and methylene blue (MB) exerts central nervous system neuroprotective effects. However, only one previous study has investigated the effectiveness of MB in a controlled cortical impact injury model of TBI. In
Toll-like 4 receptor inhibitor TAK-242 decreases neuroinflammation in rat brain frontal cortex after stress.
Garate, I; Garcia-Bueno, B; Madrigal, JL; Caso, JR; Alou, L; Gomez-Lus, ML; Leza, JC
Journal of Neuroinflammation null
Andrea K Stacy et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 43(19), 3495-3508 (2023-04-08)
Selectivity for direction of motion is a key feature of primary visual cortical neurons. Visual experience is required for direction selectivity in carnivore and primate visual cortex, but the circuit mechanisms of its formation remain incompletely understood. Here, we examined

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service