Skip to Content
Merck
All Photos(2)

Key Documents

ABN278

Sigma-Aldrich

Anti-Prospero homeobox protein 1/PROX1 Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

Prospero homeobox protein 1, Homeobox prospero-like protein PROX1, PROX-1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

mouse, rat

species reactivity (predicted by homology)

bovine (based on 100% sequence homology), fish (based on 100% sequence homology), chicken (based on 100% sequence homology), human (based on 100% sequence homology)

technique(s)

immunohistochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

mouse ... Prox1(19130)

General description

PROX1 (Prospero homeobox protein 1), the homolog of Drosophila Prospero, is a homeobox-containing transcription factor that binds and functions as a co-receptor of liver receptor homolog 1 (LRH1/NR5A2). It is a specific marker for lymphatic endothelial cells, as well as hepatoblasts and hepatocytes. Human Prox1 shares 97% amino acid sequence identity with mouse and rat Prox1.

Immunogen

KLH-conjugated linear peptide corresponding to the C-terminus of human Prospero homeobox protein 1/PROX1.

Application

Immunohistochemistry Analysis: A 1:2,000 dilution from a representative lot detected Prospero homeobox protein 1/PROX1 in mouse E16 embryo thalamus tissues (Prof. Samual Pleasure, University of California, San Francisco.).

Alexa Fluor is a registered trademark of Life Technologies, Inc.
This Anti-Prospero homeobox protein 1/PROX1 Antibody is validated for use in western blotting & IHC for the detection of Prospero homeobox protein 1/PROX1.

Quality

Evaluated by Western Blotting in HEK293 nuclear extract.

Western Blotting Analysis: A 1:500 dilution of this antibody detected Prospero homeobox protein 1/PROX1 in 10 µg of HEK293 nuclear extract.

Target description

~95 kDa observed. Uniprot describes a molecular weight at 83 kDa However, this protein may be observed between 80-110 kDa due to sumoylation and/or alternative start sites (Pan, M. R., et al. (2009). J Cell Sci. 122(Pt18):3358-3361.; Azuma, K., et al. (2011). Genes Cells. 16(11):1063-1070.).

Linkage

Replaces: AB5475

Legal Information

ALEXA FLUOR is a trademark of Life Technologies

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Charlotte M Ermine et al.
Journal of neuroscience research, 99(12), 3222-3237 (2021-10-16)
Animal modeling has played an important role in our understanding of the pathobiology of stroke. The vast majority of this research has focused on the acute phase following severe forms of stroke that result in clear behavioral deficits. Human stroke
Hidenori Ito et al.
Hippocampus, 29(7), 569-578 (2018-11-06)
The hippocampus is generally considered as a brain center for learning and memory. We have recently established an electroporation-mediated gene transfer method to investigate the development of neonatal dentate granule cells in vivo. Using this new technique, we introduced knockdown
Lu M Yang et al.
PLoS genetics, 15(7), e1008254-e1008254 (2019-07-06)
The mouse organ of Corti, housed inside the cochlea, contains hair cells and supporting cells that transduce sound into electrical signals. These cells develop in two main steps: progenitor specification followed by differentiation. Fibroblast Growth Factor (FGF) signaling is important
Charlotte M Ermine et al.
Hippocampus, 28(5), 327-337 (2018-02-13)
Key pathological features of Parkinson's Disease (PD) include the progressive degeneration of midbrain dopaminergic (DA) neurons and hindbrain noradrenergic (NA) neurons. The loss of DA neurons has been extensively studied and is the main cause of motor dysfunction. Importantly, however
In-Young Choi et al.
Frontiers in molecular neuroscience, 15, 917598-917598 (2022-07-26)
Anxiety disorder is one of the most common comorbidities in temporal lobe epilepsy (TLE), but its neurobiological mechanisms remain unclear. Here we identified a novel target, interleukin-17A (IL-17A), which can contribute to TLE-associated anxiety. Epileptic seizures were induced in 6-week-old

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service