Skip to Content
Merck
All Photos(2)

Key Documents

ABN242

Sigma-Aldrich

Anti-Pannexin-1/PANX1 Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

Pannexin-1, Innexin

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human, mouse, rat

species reactivity (predicted by homology)

gorilla (based on 100% sequence homology), chimpanzee (based on 100% sequence homology)

technique(s)

immunohistochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... PANX1(24145)

General description

Pannexin-1 (UniProt Q96RD7; also known as Innexin) is encoded by the PANX1 (also known as MRS1, PX1) gene (ORF UNQ2529/PRO6028; Gene ID 24145) in human. Pannexin 1 is a high-conductance, voltage-gated channel protein widely expressed in many organs and tissues, including the sensory systems (eye, inner ear, taste buds, and the olfactory epithelium) and both neuronal and non-neuronal cell types. Three pannexin family genes (PANX1-3) have been identified in human and mouse genomes. Pannexin proteins share structural and functional similarities and play important roles in many physiological and pathological processes, such as ATP release, Ca2+ homeostasis, synaptic neurotransmitters release, apoptosis, and immunological response. Pannexin-1 deficiency causes caspase-3 activation and apoptosis induction of hair cells in cochlear spiral limbus (SLM), resulting in hearing loss among Panx1-/- mice. Panx1 is also expressed in the retina, lens and cornea of the eye, where it plays an important role in mediating the crosstalk between immune and neuronal cells. Pannexin-1 channel opening, frequently together with the activation of purinergic receptors by extracellular ATP, is reported to have a great implication in diverse pathological conditions, including ischemia, trauma, HIV infection, inflammation, migraine, and cancer cell growth.

Specificity

Expected to react with both spliced isoforms reported by UniProt (Q96RD7-1 & Q96RD7-2).

Immunogen

Epitope: C-terminal
GST-tagged recombinant human Pannexin-1/PANX1 C-terminal fragment.

Application

Anti-Pannexin-1/PANX1 Antibody is an antibody against Pannexin-1/PANX1 for use in Western Blotting, Immunohistochemistry.
Immunohistochemistry Analysis: A 1:250 dilution from a representative lot detected Pannexin-1/PANX1 in human testis and mouse kidney tissue sections.
Research Category
Neuroscience
Research Sub Category
Signaling Neuroscience

Quality

Evaluated by Western Blotting in human cortex tissue lysate.

Western Blotting Analysis: 0.1 µg/mL of this antibody detected Pannexin-1/PANX1 in 10 µg of human cortex tissue lysate.

Target description

~55 kDa observed. 55-58 kDa (membrane glycosylated) and 43 kDa (cytosolic non-glycosylated) expected. 48.1 kDa (isoform 1) & 47.6 kDa (isoform 2) calculated.

Linkage

Replaces: AB9886

Physical form

Affinity purified
Purified rabbit polyclonal antibody in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Josip Lesko et al.
Biomedicines, 10(3) (2022-03-26)
We investigated DAB1-protein deficiency in the inner-ear development of yotari in comparison to humans and wild-type (wt) mice by immunofluorescence for the expression of connexins (Cxs) and the pannexin Panx1. The spatial and temporal dynamics of Cx26, Cx32, Cx37, Cx40
Diana Meter et al.
International journal of molecular sciences, 23(11) (2022-06-11)
The aim of this study was to determine the effects of altered ganglioside composition on the expression of Cx37, Cx40, Cx43, Cx45, and Panx1 in different kidney regions of St8sia1 gene knockout mice (St8sia1 KO) lacking the GD3 synthase enzyme.
Ivo Jeličić et al.
Biomedicines, 10(5) (2022-05-29)
Pannexins are transmembrane glycoproteins that constitute channels involved in purinergic signaling through ATP release from cells in various physiological and pathological processes. In this study, the distribution of Panx1 expression in different cell populations of healthy postnatal human kidneys and
Carolina Flores-Muñoz et al.
International journal of molecular sciences, 22(3) (2021-01-28)
Wound healing is a dynamic process required to maintain skin integrity and which relies on the precise migration of different cell types. A key molecule that regulates this process is ATP. However, the mechanisms involved in extracellular ATP management are
Carolina Flores-Muñoz et al.
Cells, 11(22) (2022-11-27)
Enhanced activity and overexpression of Pannexin 1 (Panx1) channels contribute to neuronal pathologies such as epilepsy and Alzheimer's disease (AD). The Panx1 channel ablation alters the hippocampus's glutamatergic neurotransmission, synaptic plasticity, and memory flexibility. Nevertheless, Panx1-knockout (Panx1-KO) mice still retain

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service