Skip to Content
Merck
All Photos(2)

Documents

ABN1698

Sigma-Aldrich

Anti-Aspa/Nur7 Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

Aspartoacylase, Aminoacylase-2, ACY-2, Aspa/Nur7, Anti-Aspartoacylase

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human, mouse, rat

technique(s)

immunohistochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... ASPA(443)

General description

Aspartoacylase (EC 3.5.1.15; UniProt Q8R3P0; also known as Acy-2, Aminoacylase-2, Nur-7) is encoded by the Aspa (also known as Acy2) gene (Gene ID 11484) in murine species. Aspartoacylase catalyzes the deacetylation of N-acetylaspartate (NAA) to generate free acetate in the central nervous system (CNS). Mutations in the ASPA gene cause the autosomal recessive neurodegenerative Canavan disease (CD) that results in inadequate lipid/myelin synthesis during development. Immunohistochemical staining reveals that aspartoacylase colocalizes with the oligodendrocyte marker CC1 throughout the brain, indicating its role in myelination.

Immunogen

N-terminally his-tagged full-length recombinant mouse Aspa/Nur7.

Application

Immunohistochemistry Analysis: A representative lot detected Aspa/Nur7-positive oligodendrocytes in corpus callosum/external capsule using free-floating mouse brain sections (Courtesy of Dr. John R. Moffett, Uniformed Services University of the Health Sciences).
Western Blotting Analysis: A representative lot detected Aspa/Nur7 in the cytosol, but not myelin, fraction from rat brain tissue homogenate (Madhavarao, C.N., et al. (2004). J Comp Neurol. 472(3):318-329).
Immunohistochemistry Analysis: A representative lot detected Aspa/Nur7 expression pattern in various regions of rat forebrain, including corpus callosum, cerebral cortex, hippocampal commissure (hc), fimbria, and anterior commissure (Madhavarao, C.N., et al. (2004). J Comp Neurol. 472(3):318-329).
Immunohistochemistry Analysis: A representative lot detected similar Aspa/Nur7 expression pattern as CC1 in various regions of rat forebrain, including the Purkinjie & axonal fiber layers of cerebellum, corpus callosum, as well as layer 2 of primary somatosensory cortex (Madhavarao, C.N., et al. (2004). J Comp Neurol. 472(3):318-329).
Research Category
Neuroscience
Research Sub Category
Developmental Signaling
This Anti-Aspa/Nur7 Antibody is validated for use in Western Blotting, Immunohistochemistry for the detection of Aspa/Nur7.

Quality

Evaluated by Western Blotting in rat brain tissue lysate.

Western Blotting Analysis: 1.0 µg/mL of this antibody detected Aspa/Nur7 in 10 µg of rat brain tissue lysate.

Target description

~37 kDa observed

Physical form

Protein G Purified
Purified rabbit polyclonal antibody in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yun-Yun Hu et al.
Frontiers in psychiatry, 14, 1186073-1186073 (2023-07-06)
Social interaction is a fundamental human need. Social isolation (SI) can have negative effects on both emotional and cognitive function. However, it is currently unclear how age and the duration of SI affect emotion and recognition function. In addition, there
Lillian M Toomey et al.
Scientific reports, 11(1), 22594-22594 (2021-11-21)
Cuprizone is a copper-chelating agent that induces pathology similar to that within some multiple sclerosis (MS) lesions. The reliability and reproducibility of cuprizone for inducing demyelinating disease pathology depends on the animals ingesting consistent doses of cuprizone. Cuprizone-containing pelleted feed
Zhixin Lei et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 40(33), 6444-6456 (2020-07-15)
Previous studies demonstrate that activation of pancreatic ER kinase (PERK) protects oligodendrocytes against inflammation in the experimental autoimmune encephalomyelitis (EAE) model of multiple sclerosis (MS). Interestingly, data indicate that the cytoprotective effects of PERK activation on oligodendrocytes during EAE are
Simon Pan et al.
Nature neuroscience, 23(4), 487-499 (2020-02-12)
Experience-dependent myelination is hypothesized to shape neural circuit function and subsequent behavioral output. Using a contextual fear memory task in mice, we demonstrate that fear learning induces oligodendrocyte precursor cells to proliferate and differentiate into myelinating oligodendrocytes in the medial
P Chaudhary et al.
Journal of neuroimmunology, 352, 577468-577468 (2021-01-11)
We have previously demonstrated that thyromimetics stimulate oligodendrocyte precursor cell differentiation and promote remyelination in murine demyelination models. We investigated whether a thyroid receptor-beta selective thyromimetic, sobetirome (Sob), and its CNS-targeted prodrug, Sob-AM2, could prevent myelin and axonal degeneration in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service