Skip to Content
Merck

ABF23

Anti-NLRP3 Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

NACHT, LRR and PYD domains-containing protein 3, Angiotensin/vasopressin receptor AII/AVP-like, Caterpiller protein 1.1, CLR1.1, Cold autoinflammatory syndrome 1 protein, Cryopyrin, PYRIN-containing APAF1-like protein 1, NLRP3

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
eCl@ss:
32160702
Conjugate:
unconjugated
Clone:
polyclonal
Application:
IHC, WB
Citations:
2
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

rabbit

Quality Segment

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human

technique(s)

immunohistochemistry: suitable (paraffin), western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... NLRP3(114548)

General description

NLRPs (Nucleotide-binding oligomerization domain, Leucine rich Repeat and Pyrin domain containing Proteins) are members of NLR (Nod-like receptors) protein family. NRLP genes are known to play important roles in both the mammalian reproductive system and the innate immune system. NLRP3 may function as an inducer of apoptosis, activator of NF-kappa-B signaling and activates caspase-1 in response to triggers such as bacterial or viral infection. NLRP3 defects are linked to familial cold autoinflammatory syndrome type 1 (FCAS1), Muckle-Wells syndrome (MWS) and chronic infantile neurologic cutaneous and articular syndrome (CINCA). NLRP3 is highly expressed in blood leukocytes, polymorphonuclear cells, osteoblasts and chondrocytes.
~112 kDa and ~75 kDa observed. Five isoforms are known to exist at 118 kDa, 112 kDa, 112 kDa, 106 kDa, and 84 kDa This protein has been observed at ~75 kDa in some cell lysates (Kummer, J. A., et al. (2007). J Histochem Cytochem. 55(5):443-452.). Uncharacterized bands may be observed in some cell lysates.

Immunogen

Epitope: DAPIN domain
KLH-conjugated linear peptide corresponding to the DAPIN domain of human NLRP3.

Application

Detect NLRP3 using this rabbit polyclonal antibody, Anti-NLRP3 Antibody validated for use in western blotting & IHC (Paraffin).
Immunohistochemistry Analysis: A 1:4,000 dilution from a representative lot detected NLRP3 in human leukemia bone marrow and human tonsil tissue lysate.
Research Category
Inflammation & Immunology
Research Sub Category
Immunological Signaling

Biochem/physiol Actions

Other homologies: Porcine (80% sequence homology).
This antibody recognizes the DAPIN domain of NLRP3.

Physical form

Affinity purified
Purified rabbit polyclonal in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Preparation Note

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
THP-1 cell lysate
Evaluated by Western Blotting in THP-1 cell lysate.

Western Blotting Analysis: 1 µg/mL of this antibody detected NLRP3 in 10 µg of THP-1 cell lysate.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.


Still not finding the right product?

Try our Product Selector Tool to narrow your options


Storage Class

12 - Non Combustible Liquids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library