Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
UNSPSC Code:
12352203
NACRES:
NA.41
eCl@ss:
32160702
Conjugate:
unconjugated
Clone:
polyclonal
Application:
IF, WB
Citations:
-
biological source
rabbit
Quality Segment
conjugate
unconjugated
antibody form
affinity isolated antibody
antibody product type
primary antibodies
clone
polyclonal
species reactivity
mouse
packaging
antibody small pack of 25 μL
technique(s)
immunofluorescence: suitable, western blot: suitable
isotype
IgG
NCBI accession no.
UniProt accession no.
shipped in
ambient
target post-translational modification
unmodified
Gene Information
mouse ... Fcgrt(14132)
General description
IgG receptor FcRn large subunit p51 (UniProt: Q61559; also known as FcRn, IgG Fc fragment receptor transporter alpha chain, Neonatal Fc receptor) is encoded by the Fcgrt (also known as Fcrn) gene (Gene ID: 14132) in murine species. FcRn is a single-pass type I membrane protein that binds to the Fc region of monomeric immunoglobulins gamma. FcRn complex consist of two subunits: p51 and p14 and contains one Ig-like C1-type (immunoglobulin-like) domain of 88 amino acids. FcRn mediates the selective uptake of IgG from milk and helps newborn animals to acquire passive immunity. In rodents, FcRn functions effectively in the neonatal period when it transports maternally derived IgG in ingested milk across the epithelial-cell layer of the proximal small intestine. However, in humans it transports maternal antibody to the fetus across the placenta. In rodent FcRn-IgG complexes are transcytosed across the intestinal epithelium and IgG is released from FcRn into blood or tissue fluids. FcRn also protects IgG from degradation providing long half-life of this class of antibody in the serum. (Ref.: Roopenian,, DC and Shreeram,, A (2007). Nat. Rev. Immunol. 7(9); 715-725).
~53 kDa observed; 40.09 kDa calculated. Uncharacterized bands may be observed in some lysate(s).
Immunogen
Epitope: C-terminus
GST-tagged recombinant fragment corresponding to 44 amino acids from the C-terminal region of murine IgG receptor FcRn large subunit p51.
Application
Immunofluorescence Analysis: A representative lot detected FcRn (FCGRT) in mouse uterus and vagina (Li, Z., et. al. (2011). Proc Natl Acad Sci USA. 108(11):4388-93).
Western Blotting Analysis: A representative lot detected FcRn (FCGRT) in mouse liver, mouse uterus, and vagina tissue lysate (Li, Z., et. al. (2011). Proc Natl Acad Sci USA. 108(11):4388-93).
Western Blotting Analysis: A representative lot detected FcRn (FCGRT) in mouse liver, mouse uterus, and vagina tissue lysate (Li, Z., et. al. (2011). Proc Natl Acad Sci USA. 108(11):4388-93).
Research Category
Inflammation & Immunology
Inflammation & Immunology
This rabbit polyclonal Anti-FcRn (FCGRT) antibody, Cat. No. ABF191, detects IgG receptor FcRn large subunit p51. It has been tested for use in Immunofluorescence and Western Blotting.
Biochem/physiol Actions
This polyclonal antibody detects murine IgG receptor FcRn. It targets an epitope within 44 amino acids from the C-terminal region.
Physical form
Affinity Purified
Format: Purified
Purified rabbit polyclonal antibody in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.
Preparation Note
Stable for 1 year at 2-8°C from date of receipt.
Analysis Note
Evaluated by Western Blotting in mouse liver tissue lysate.
Western Blotting Analysis: A 1:500 dilution of this antibody detected FcRn (FCGRT) in 10 µg of mouse liver tissue lysate.
Western Blotting Analysis: A 1:500 dilution of this antibody detected FcRn (FCGRT) in 10 µg of mouse liver tissue lysate.
Other Notes
Concentration: Please refer to lot specific datasheet.
Disclaimer
Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.
Still not finding the right product?
Try our Product Selector Tool to narrow your options
Storage Class
12 - Non Combustible Liquids
wgk
WGK 1
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.