Skip to Content
Merck
All Photos(4)

Key Documents

ABE44

Sigma-Aldrich

Anti-trimethyl Histone H3 (Lys27) Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

H3K27me3, Histone H3 (tri methyl K27), H3 histone family, member T, histone 3, H3, histone cluster 3, H3, H3K27me3

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

mouse, human

technique(s)

dot blot: suitable
immunocytochemistry: suitable
inhibition assay: suitable (peptide)
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

trimethylation (Lys27)

Gene Information

human ... HIST1H3F(8968)

General description

Histone H3 is a core component of nucleosome. Nucleosomes wrap and compact DNA into chromatin, limiting DNA accessibility to the cellular machineries which require DNA as a template. Histones thereby play a central role in transcription regulation, DNA repair, DNA replication and chromosomal stability. DNA accessibility is regulated via a complex set of post-translational modifications of histones, also called histone code, and nucleosome remodeling. Histone H3 lysine 27 methylation (H3K27) is associated with heterochromatin formation and transcriptional repression.

Specificity

Broad species cross-reactivity expected.
This antibody recognizes Histone H3 methylated at Lys27.

Immunogen

Branch peptide corresponding to human Histone H3 methylated at Lys27.
Epitope: Lys27

Application

Anti-trimethyl Histone H3 (Lys27) Antibody is a rabbit polyclonal antibody for detection of Histone H3 trimethylated on lysine 27( H3K27me3). This purified polyclonal is specificity verified by dot blot (DB), published in peer reviewed journals and validated in WB, ICC, PIA.
Immunocytochemistry Analysis: 1:500 dilution from a representative lot detected Histone H3 in NIH/3T3 cells.
Peptide Inhibition Assay: This antibody peptide blocked on HeLa cell extracts.
Research Category
Epigenetics & Nuclear Function
Research Sub Category
Histones

Quality

Evaluated by Western Blot in HeLa acid extract and recombinant Histone H3.

Western Blot Analysis: 0.1 µg/ml of this antibody detected histone H3 in 10 µg of HeLa acid extract .

Target description

~ 17 kDa

Physical form

Affinity purified
Purified rabbit polyclonal in buffer containing 0.1 M Tris-Glycine (pH 7.4, 150 mM NaCl) with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
HeLa acid extract and recombinant Histone H3

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chromatin signature of widespread monoallelic expression.
Nag, A; Savova, V; Fung, HL; Miron, A; Yuan, GC; Zhang, K; Gimelbrant, AA
eLife null
Development of multiple cell-based assays for the detection of histone H3 Lys27 trimethylation (H3K27me3).
Qian, J; Lu, L; Wu, J; Ma, H
Assay and Drug Development Technologies null
Chromatin Signature Identifies Monoallelic Gene Expression Across Mammalian Cell Types.
Nag, A; Vigneau, S; Savova, V; Zwemer, LM; Gimelbrant, AA
G3 (Bethesda, Md.) null
Hao-Kang Li et al.
Scientific reports, 6, 28637-28637 (2016-06-28)
Studies indicate that the presence of cancer stem cells (CSCs) is responsible for poor prognosis of hepatocellular carcinoma (HCC) patients. In this study, the functional role of DDX3 in regulation of hepatic CSCs was investigated. Our results demonstrated that reduced
Jung Kim et al.
Cell reports, 25(10), 2808-2820 (2018-12-06)
Enhancer of Zeste 2 (EZH2) is the enzymatic subunit of Polycomb Repressive Complex 2 (PRC2), which catalyzes histone H3 lysine 27 trimethylation (H3K27me3) at target promoters for gene silencing. Here, we report that EZH2 activates androgen receptor (AR) gene transcription

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service