Skip to Content
Merck

Skip To

ABE208

Anti-β-Catenin Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

catenin (cadherin-associated protein), beta 1, 88kDa, catenin (cadherin-associated protein), beta 1 (88kD)

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
eCl@ss:
32160702
Conjugate:
unconjugated
Clone:
polyclonal
Application:
ICC, IP, WB
Citations:
7
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

rabbit

Quality Segment

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

mouse, human, rat, horse

species reactivity (predicted by homology)

ox (based on 100% sequence homology), equine (based on 100% sequence homology)

technique(s)

immunocytochemistry: suitable, immunoprecipitation (IP): suitable, western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

General description

β-catenin is the founding member of the β -catenin family. This protein is comprised of 12 ARM repeats and is considered to have dual function depending on its localization. Cytoplasmic β -catenin is a critical component of E-cadherin/catening adhesion complexes and is involved in regulating cadherin-dependent cell-cell interactions.When localized to the nucleus, β -catenin serves a regulatory role by binding with transcription factors from the TCF/LEF family, activating transcription. Expression of β -catenin has been reported in a variety of hair follicle cells such as cells found in the inner and outer portions of the root sheaths, and the basal and peripheral matrix cells. It has also been observed in colon tissue. Defects in expression have been linked to the development of several tumors and cancers such as; colorectal cancer (CRC), medulloblastoma (MDB), ovarian cancer (OC), and pilomatrixoma (PTR).
~86kDa observed.

Immunogen

Epitope: Consensus GSK3 phosphorylation site of human β-Catenin
KLH-conjugated linear peptide corresponding to the consensus GSK3 phosphorylation site of human β-Catenin.

Application

Anti-β-Catenin Antibody is a Rabbit Polyclonal Antibody for detection of β-Catenin also known as catenin (cadherin-associated protein) beta 1 (88kD) & has been validated in WB, ICC & IP.
Immunoprecipitation Analysis: 10 µg from a representative lot immunoprecipitated β-Catenin from A431 lysate.

Immunocytochemistry Analysis: 1:500 dilution from a representative lot detected β-Catenin in NIH/3T3 and A431 cells.
Research Category
Epigenetics & Nuclear Function
Research Sub Category
Transcription Factors

Biochem/physiol Actions

Antibody recognizes β-Catenin at the N-terminus.

Physical form

Affinity purified
Purified rabbit polyclonal in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Preparation Note

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
A431 cell lysate
Evaluated by Western Blot in A431 cell lysate.

Western Blot Analysis: 0.05 µg/mL of this antibody detected β-Catenin on 10 µg of A431 cell lysate.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.
Replaces: 04-1002

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
MAB2081SAB5700055SAB4500545
description

from rabbit, purified by affinity chromatography

description

clone 5H10, Chemicon®, from mouse

description

-

description

affinity isolated antibody

conjugate

unconjugated

conjugate

unconjugated

conjugate

unconjugated

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody form

purified immunoglobulin

antibody form

affinity isolated antibody

antibody form

affinity isolated antibody

Quality Level

100

Quality Level

100

Quality Level

100

Quality Level

100

biological source

rabbit

biological source

mouse

biological source

rabbit

biological source

rabbit

technique(s)

immunocytochemistry: suitable, western blot: suitable, immunoprecipitation (IP): suitable

technique(s)

immunocytochemistry: suitable, immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable, immunoprecipitation (IP): suitable, western blot: suitable

technique(s)

immunohistochemistry: 1:50-1:200, immunoprecipitation (IP): 1:50-1:200, western blot: 1:500-1:2000

technique(s)

ELISA: 1:40000, western blot: 1:500-1:1000

Gene Information

horse ... Ctnnb1(100055241)
human ... CTNNB1(1499)
mouse ... Ctnnb1(539003), Ctnnb1(12387)
rat ... Ctnnb1(84353)

Gene Information

human ... CTNNB1(1499)
rat ... Ctnnb1(84353)

Gene Information

human ... CTNNB1(1499)

Gene Information

human ... CTNNB1(1499)


Still not finding the right product?

Explore all of our products under Anti-β-Catenin Antibody

or

Try our Product Selector Tool to narrow your options


Storage Class

12 - Non Combustible Liquids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters