Skip to Content
Merck

ABE1889

Anti-ZMYND11 Antibody

serum, from rabbit

Synonym(s):

Zinc finger MYND domain-containing protein 11

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
eCl@ss:
32160702
Conjugate:
unconjugated
Clone:
polyclonal
Application:
ChIP, WB
Citations:
-
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

rabbit

Quality Segment

conjugate

unconjugated

antibody form

serum

antibody product type

primary antibodies

clone

polyclonal

species reactivity

human

species reactivity (predicted by homology)

mouse (based on 100% sequence homology)

technique(s)

ChIP: suitable (ChIP-seq), western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

mouse ... Zmynd11(66505)

General description

Zinc finger MYND domain-containing protein 11 (UniProt Q8R5C8) is encoded by the Zmynd11 gene (Gene ID 66505) in murine species. ZMYND11/BS69 functions as a co-repressor of adenovirus E1A and cellular transcription factors including the c-Myb and ETS-2 oncoproteins. ZMYND11 contains several histone reader motifs, including a plant homeodomain (PHD), a bromodomain, and a PWWP domain. Lys36-trimethylated histone H3 (H3K36me3) is deposited onto the nucleosomes in the transcribed regions after RNA polymerase II elongation, and ZMYND11 is found to specifically recognize H3K36me3 and regulate RNA polymerase II elongation. Binding study showed that the combined PHD-bromo-PWWP (PBP) module mediates ZMYND11 recognition of H3K36me3 and PWWP domain alone displays little to no affinity toward H3K36me, H3K36me2, or unmethylated H3K36. ZMYND11 is a tumor suppressor that functions as an unconventional transcription co-repressor by modulating RNA polymerase II at the elongation stage. Low expression levels of ZMYND11 in breast cancer patients correlate with poor prognosis. Consistently, ZMYND11 overexpression suppresses cancer cell growth in vitro and tumor formation in mice in vivo.
~70 kDa observed. 70.96 kDa (isoform 1), 64.43 kDa (isoform 2), 66.59 kDa (isoform 3), 60.05 kDa (isoform 4), 60.87 kDa (isoform 5), and 66.52 kDa (isoform 6) calculated.

Immunogen

Epitope: C-terminal region.
His-tagged recombinant mouse ZMYND11 C-terminal fragment.

Application

Detect ZMYND11 using this Anti-ZMYND11 Antibody validated for use in Western Blotting, Chromatin Immunoprecipitation (ChIP), ChIP-seq.
Research Category
Epigenetics & Nuclear Function
Research Sub Category
Chromatin Biology
Western Blotting Analysis: A 1:1,000 dilution from a representative lot detected a downregulated ZMYND11 level in human osteosarcoma U2OS cell nuclear extracts following ZMYND11 shRNA treatment (Courtesy of Dr. Hong Wen, UT M.D. Anderson Cancer Center, TX).
Chromatin Immunoprecipitation Analysis: A representative lot detected ZMYND11 occupancy of c-Myc gene by ChIP using human osteosarcoma U2OS cell nuclear extracts. ZMYND11 shRNA treatment greatly reduced the c-Myc gene fragments in ChIP (Courtesy of Dr. Hong Wen, UT M.D. Anderson Cancer Center, TX).
Chromatin Immunoprecipitation (ChIP) Analysis: A representative lot detected ZMYND11 chromatin occupancy by ChIP using chromatin preparations from human osteosarcoma U2OS cells. Decreasing H3K36me3 level by SETD2 knockdown reduced ZMYND11 target genes association (Wen, H., et al. (2014). Nature. 508(7495):263-268).
ChIP-seq Analysis: A representative lot detected ZMYND11-targeted chromatin sites by a genome-wide ChIP-seq analysis using chromatin preparations from human osteosarcoma U2OS cells. The ZMYND11 occupied sites are highly enriched in introns and exons, but not promoters of targeted genes (Wen, H., et al. (2014). Nature. 508(7495):263-268).
Western Blotting Analysis: A representative lot detected exogenously expressed human BS69 protein in yeast and QT-6 quail fibroblasts (Ladendorff, N.E., et al. (2001). Oncogene. 20(1):125-132).

Biochem/physiol Actions

This antiserum detects two immunoreactive bands by Western blotting analysis of U2OS nuclear extract, ZMYND11 shRNA treatment specifically abolished the ~70 kDa band, but not the ~60 kDa band. The detection of the non-ZMYND11 band by Western blotting does not affect this antiserum′s performance in chromatin immunoprecipitation (ChIP) application. ZMYND11-knockdown by shRNA treatment prior to ChIP with this antiserum abolished the DNA fragments present in the immunoprecipitates from non-shRNA treated cells. In addition, ChIP profile obtained with this antiserum closely resembles that obtained using an anti-FLAG antbody and cells expressing FLAG-tagged ZMYND11.

Physical form

Rabbit polyclonal serum with 0.05% sodium azide.
Unpurified

Preparation Note

Stable for 1 year at -20°C from date of receipt.
Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.

Analysis Note

Evaluated by Western Blotting in U2OS nuclear extract.

Western Blotting Analysis: A 1:500 dilution of this antibody detected ZMYND11 in 20 µg of U2OS nuclear extract.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.


Still not finding the right product?

Try our Product Selector Tool to narrow your options


Storage Class

12 - Non Combustible Liquids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library