Skip to Content
Merck

ABC932

Anti-IFT88 Antibody

serum, from rabbit

Synonym(s):

Intraflagellar transport protein 88 homolog, Polaris, Recessive polycystic kidney disease protein Tg737, Tetratricopeptide repeat protein 10, TgN(Imorpk)737Rpw, TPR repeat protein 10

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
eCl@ss:
32160702
Conjugate:
unconjugated
Clone:
polyclonal
Application:
EM, ICC, IF, WB
Citations:
1
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

rabbit

Quality Segment

conjugate

unconjugated

antibody form

serum

antibody product type

primary antibodies

clone

polyclonal

species reactivity

rat, human, mouse

species reactivity (predicted by homology)

bovine (based on 100% sequence homology)

technique(s)

electron microscopy: suitable, immunocytochemistry: suitable, immunofluorescence: suitable, western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

bovine ... Ift88(514177)
human ... IFT88(8100)
mouse ... Ift88(21821)
rat ... Ift88(305918)

General description

Intraflagellar transport protein 88 homolog (UniProt Q61371; also known as Polaris, Recessive polycystic kidney disease protein Tg737, Tetratricopeptide repeat protein 10, TgN(Imorpk)737Rpw, TPR repeat protein 10) is encoded by the Ift88 (also known as Tg737, Tg737Rpw, TgN737Rpw, Ttc10) gene (Gene ID 21821) in murine species. The primary cilium is a microtubule-based mechano- and chemo-sensory organelle that coordinates an array of cellular pathways during development and in tissue homeostasis, including Hedgehog (Hh), PDGFRα and Wnt signaling. Primary cilium is assembled and maintained via a process termed intraflagellar transport (IFT) in mammalian cells during growth arrest. Ift88/Polaris/Tg737 is a subunit of the IFT particle complex B required for the assembly of functional IFT. Defects in primary cilium assembly lead to severe developmental diseases and disorders referred to as ciliopathies. Polycystic kidney disease (PKD) is one of the first diseases identified to be linked to dysfunctional primary cilia. PKD was originally identified in the Oak Ridge Polycystic Kidney (ORPK) mouse harboring mutated Ift88 gene (Ift88orpk or Ift88Tg737NRpw). Ift88/Polaris/Tg737 is commonly used as a primary cilium marker.
~95 kDa observed. 92.99 kDa calculated.

Immunogen

Epitope: Region between TPR repeats 3 & 4.
Linear peptide corresponding to a mouse IFT88 sequence from the region between TPR repeats 3 & 4.

Application

Detect IFT88 using this Anti-IFT88 Antibody validated for use in Western Blotting, Immunocytochemistry, Electron Microscopy, Immunofluorescence.
Immunocytochemistry Analysis: An 1:500 dilution from a representative lot immunostained primary cilia of paraformaldehyde-fixed mouse kidney tubule cells (Courtesy of Dr. Bradley K. Yoder, University of Alabama Birmingham, USA).
Western Blotting Analysis: A representative lot detected a greatly reduced Tg737/Polaris expression in lysate from E11.5 mouse embryo limbs with conditional mesenchymal Ift88-knockout (prx1cre;Ift88fl/n) when compared with lysate from a wild-type littermate (Haycraft, C.J., et al. (2007). Development. 134(2):307-316).
Immunocytochemistry Analysis: A representative lot detected IFT88 (Tg737/Polaris) cellular localization in paraformaldehyde-fixed rat and murine sperm cells, rat round spermatids, as well as elongating spermatids from Ift88 mutant mice (Kierszenbaum, A.L., et al. (2011). Dev. Dyn. 240(3):723-736).
Immunocytochemistry Analysis: A representative lot detected Tg737/Polaris immunoreactivity associated with the cilium of cultured limb bud cells isolated from wild-type mouse E11.5 embryos by fluorescent immunocytochemistry, while cilia were absent in cells from Tg737Δ2–3β-gal embryos (Haycraft, C.J., et al. (2005). PLoS Genet. 1(4):e53).
Electron Microscopy Analysis: A representative lot detected IFT88 (Tg737/Polaris) localization in rat round spermatids and fractionated manchettes (Kierszenbaum, A.L., et al. (2011). Dev. Dyn. 240(3):723-736).
Immunofluorescence Analysis: A representative lot detected polaris immunoreactivity in both dental epithelium (DE) and dental mesenchyme (DM) of lower molar frontal sections from E13.5 and E16.5 embryonic mice (Ohazama, A., et al. (2009). Development. 136(6):897-903).
Immunofluorescence Analysis: A representative lot immunostained primary cilium on epithelial and mesenchymal cells of the skin and hair follicle using OCT-embedded frozen skin sections from both mouse embryos and adult mice (Lehman, J.M., et al. (2009). J. Invest. Dermatol. 129(2):438-448).
Immunofluorescence Analysis: A representative lot detected Ift88 (Tg737/Polaris) immunoreactivity enriched at the base and tip of the hair cell (HC) kinocilia in the ciliary axonemes by fluorescent immunohistochemistry using 4% paraformaldehyde-fixed inner ear cochlear whole mounts prepared from mouse E14.5 embryos (Jones, C., et al. (2008). Nat. Genet. 40(1):69-77).
Immunofluorescence Analysis: A representative lot immunostained cilium in mesenchymal and ectodermal cells in formaldehye-fixed, OCT-embedded frozen limb bud sections from mouse E11.5 embryos by fluorescent immunohistochemistry. Conditional mesenchymal Ift88-knockout (prx1cre;Ift88fl/n) selectively abolished Tg737/Polaris immunoreactivity in the mesenchyme, but not the ectoderm layer (Haycraft, C.J., et al. (2007). Development. 134(2):307-316).
Immunofluorescence Analysis: A representative lot detected Tg737/Polaris immunoreactivity at the base and distal tip of cilium in mesenchymal and ectodermal cells in OCT-embedded frozen limb bud sections from mouse E10.5 embryos by fluorescent immunohistochemistry (Haycraft, C.J., et al. (2005). PLoS Genet. 1(4):e53).
Research Category
Apoptosis & Cancer
Research Sub Category
Developmental Signaling

Biochem/physiol Actions

This antiserum stains primary cilia in human, mouse, and rat cells by fluorescent immunohistochemistry and immunocytochemistry.

Physical form

Rabbit polyclonal serum with 0.05% sodium azide.
Unpurified

Preparation Note

Stable for 1 year at -20°C from date of receipt.
Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.

Analysis Note

Evaluated by Western Blotting in mouse kidney tissue lysate.

Western Blotting Analysis: A 1:200 dilution of this antibody detected IFT88 in 50 µg of mouse kidney tissue lysate.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.


Still not finding the right product?

Try our Product Selector Tool to narrow your options


Storage Class

12 - Non Combustible Liquids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library