Select a Size
| Size/SKU | Availability | Price |
|---|
About This Item
biological source
rabbit
Quality Segment
conjugate
unconjugated
antibody form
affinity isolated antibody
antibody product type
primary antibodies
clone
polyclonal
species reactivity
rat, mouse
technique(s)
immunocytochemistry: suitable, immunohistochemistry: suitable, immunoprecipitation (IP): suitable, western blot: suitable
isotype
IgG
NCBI accession no.
UniProt accession no.
shipped in
ambient
target post-translational modification
unmodified
Gene Information
rat ... Slc5A5(114613)
General description
Immunogen
Application
Apoptosis & Cancer
Immunocytochemistry Analysis: A representative lot detected Sodium-Iodine Symporter (NIS) C-term in Immunocytochemistry applications (Levy, O., et. al. (1997). Proc Natl Acad Sci USA. 94(11):5568-73; De La Vieja, A., et. al. (2004). J Cell Sci. 117(Pt 5):677-87).
Immunohistochemistry Analysis: A representative lot detected Sodium-Iodine Symporter (NIS) C-term in rat thyroid, mammary gland from lactating rat (Tazebay, U.H., et. al. (2000). Nat Med. 6(8):871-8).
Western Blotting Analysis: A representative lot detected Sodium-Iodine Symporter (NIS) C-term in Western Blotting applications (Levy, O., et. al. (1997). Proc Natl Acad Sci USA. 94(11):5568-73; Tazebay, U.H., et. al. (2000). Nat Med. 6(8):871-8; Levy, O., et. al. (1998). J Biol Chem. 273(35):22657-63; De La Vieja, A., et. al. (2004). J Cell Sci. 117(Pt 5):677-87).
Immunoprecipitation Analysis: A representative lot detected Sodium-Iodine Symporter (NIS) C-term in Immunoprecipitation applications (De La Vieja, A., et. al. (2004). J Cell Sci. 117(Pt 5):677-87; Levy, O., et. al. (1998). J Biol Chem. 273(35):22657-63).
Biochem/physiol Actions
Physical form
Preparation Note
Analysis Note
Western Blotting Analysis: 1.5 µg/mL of this antibody detected Sodium-Iodine Symporter (NIS) C-term in 10 µg of rat thyroid tissue lysate.
Other Notes
Disclaimer
1 of 1
This Item | |||
|---|---|---|---|
| Quality Level 100 | Quality Level 100 | Quality Level - | Quality Level 100 |
| biological source rabbit | biological source rabbit | biological source mouse | biological source mouse |
| conjugate unconjugated | conjugate unconjugated | conjugate unconjugated | conjugate unconjugated |
| antibody form affinity isolated antibody | antibody form affinity isolated antibody | antibody form purified immunoglobulin | antibody form purified immunoglobulin |
| UniProt accession no. | UniProt accession no. | UniProt accession no. | UniProt accession no. |
| Gene Information rat ... Slc5A5(114613) | Gene Information human ... SLC5A5(6528) | Gene Information human ... SLC5A5(6528) | Gene Information human ... SLC5A5(6528) |
Still not finding the right product?
Try our Product Selector Tool to narrow your options
Storage Class
12 - Non Combustible Liquids
wgk
WGK 1
flash_point_f
Not applicable
flash_point_c
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.



