Skip to Content
Merck
All Photos(1)

Key Documents

AB9882

Sigma-Aldrich

Anti-Huntingtin-Interacting Protein 1 Related (HIP1R) Antibody

Chemicon®, from rabbit

Synonym(s):

HIP1R

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41
clone:
polyclonal
application:
ICC
WB
species reactivity:
mouse, rat, human
technique(s):
immunocytochemistry: suitable
western blot: suitable
citations:
6

biological source

rabbit

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

polyclonal

species reactivity

mouse, rat, human

manufacturer/tradename

Chemicon®

technique(s)

immunocytochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... HIP1R(9026)

General description

Huntingtin-Interacting protein-1 related (HIP1R) is an actin-binding protein that is considered an element of the clathrin-coated pits and vesicles and polymerizes actin. HIP1R is essential for normal actin organization and endocytosis. HIP1R is equipped with a epsin N-terminal homology domain which allows it to bind 3-phosphorylated inositol lipids, which allows HIP1R to function as a tyrosine kinase stabilizing receptor and plays a role in overall cell growth and survival.

Specificity

Huntingtin-interacting protein 1 related (HIP1R). By Western blot the antibody recognizes a band at ~116 kDa on P2 microsomal fraction from embryonic rat brain.

Immunogen

Synthetic peptide

Application

Detect Huntingtin-Interacting Protein 1 Related (HIP1R) using this Anti-Huntingtin-Interacting Protein 1 Related (HIP1R) Antibody validated for use in WB & IC.
Research Category
Neuroscience
Research Sub Category
Neurodegenerative Diseases

Synapse & Synaptic Biology
Western Blot: a 1:500-1:2,000 dilution of this antibody detected Huntingtin-interacting protein 1 related (HIP1R) in 10 µg of rat brain extract and mouse brain post-nuclear supernatant.A 1:500-1:2,000 dilution of this antibody detected Huntingtin-interacting protein 1 related (HIP1R) in 10 µg of rat brain extract and mouse brain post-nuclear supernatant.

Immunocytochemistry: a previous lot of this antibody was used by an independent laboratory for IC of human cells. (Poupon, V., et al. (2008). PNAS. 105(1): 168-173.)

Optimal working dilutions must be determined by end user.

Target description

~116 kDa

Physical form

Format: Purified
Liquid

Storage and Stability

Maintain at -20°C in undiluted aliquots for up to 6 months after date of receipt. Avoid repeated freeze/thaw cycles.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M A Rice et al.
Oncogene, 35(36), 4752-4761 (2016-02-24)
Deregulation of microRNAs (miRs) contributes to progression and metastasis of prostate and other cancers. miR-23b and -27b, encoded in the same miR cluster (miR-23b/-27b), are downregulated in human metastatic prostate cancer compared with primary tumors and benign tissue. Expression of
Sophia R Majeed et al.
Nature communications, 5, 3891-3891 (2014-05-24)
The clathrin light chain (CLC) subunits participate in several membrane traffic pathways involving both clathrin and actin, through binding the actin-organizing huntingtin-interacting proteins (Hip). However, CLCs are dispensable for clathrin-mediated endocytosis of many cargoes. Here we observe that CLC depletion
Qian Yang et al.
Frontiers in molecular neuroscience, 11, 447-447 (2018-12-24)
Huntington-interacting protein 1-related protein (HIP1R) was identified on the basis of its structural homology with HIP1. Based on its domain structure, HIP1R is a putative endocytosis-related protein. Our previous study had shown that knockdown of HIP1R induces a dramatic decrease
Markus Mukenhirn et al.
Cells, 10(2) (2021-03-07)
One of the most fundamental processes of the cell is the uptake of molecules from the surrounding environment. Clathrin-mediated endocytosis (CME) is the best-described uptake pathway and regulates nutrient uptake, protein and lipid turnover at the plasma membrane (PM), cell
Mian Cao et al.
Neuron, 93(4), 882-896 (2017-02-24)
Synaptojanin 1 (SJ1) is a major presynaptic phosphatase that couples synaptic vesicle endocytosis to the dephosphorylation of PI(4,5)P2, a reaction needed for the shedding of endocytic factors from their membranes. While the role of SJ1's 5-phosphatase module in this process

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service