Skip to Content

AB9574

Anti-BACE2 Antibody

serum, Chemicon®

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
eCl@ss:
32160702
Conjugate:
unconjugated
Clone:
polyclonal
Application:
IP, WB
Citations:
1
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

rabbit

Quality Segment

conjugate

unconjugated

antibody form

serum

antibody product type

primary antibodies

clone

polyclonal

species reactivity

mouse, rat, human

manufacturer/tradename

Chemicon®

technique(s)

immunoprecipitation (IP): suitable, western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... BACE2(25825)

Immunogen

Synthetic peptide from the C-terminus of human BACE2.

Application

Research Category
Neuroscience
Research Sub Category
Neurodegenerative Diseases
This Anti-BACE2 Antibody is validated for use in IP, WB for the detection of BACE2.
Western blot: 1:2,000. The antibody recognizes the ~64 kDa BACE2 protein in HEK293, SHSy5y, N2A and PC12 cell lysate.

Immunoprecipitation

Optimal working dilutions must be determined by the end user.

Biochem/physiol Actions

The antibody recognizes the BACE2 protein.

Physical form

Rabbit serum. Liquid.

Preparation Note

Maintain at -20°C in undiluted aliquots for up to 6 months after date of receipt. Avoid repeated freeze/thaw cycles.

Other Notes

Replaces: AB5834

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.


Still not finding the right product?

Try our Product Selector Tool to narrow your options


Storage Class

11 - Combustible Solids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library