Skip to Content
Merck
All Photos(1)

Key Documents

AB9348

Sigma-Aldrich

Anti-Myelin Basic Protein Antibody

CHEMICON®, chicken polyclonal

Synonym(s):

Anti-C76307, Anti-Hmbpr, Anti-R75289, Anti-golli-mbp, Anti-jve, Anti-mld, Anti-shi

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

Product Name

Anti-Myelin Basic Protein Antibody, Chemicon®, from chicken

biological source

chicken

Quality Level

antibody form

affinity purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human, mouse

manufacturer/tradename

Chemicon®

technique(s)

immunocytochemistry: suitable
immunohistochemistry: suitable

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... MBP(4155)
mouse ... Mbp(17196)

Specificity

Recognizes Myelin Basic Protein (MBP).

Immunogen

Synthetic peptide from human/mouse MBP.

Application

Anti-Myelin Basic Protein Antibody detects level of Myelin Basic Protein & has been published & validated for use in IC, IH.
Immunohistochemistry: 1:50-1:100 using 2% paraformaldehyde fixed tissues.

Immunocytochemistry: 1:50-1:100 using 2% paraformaldehyde fixed cells.

Optimal working dilutions must be determined by the end user.
Research Category
Neuroscience
Research Sub Category
Neuronal & Glial Markers

Neurochemistry & Neurotrophins

Physical form

Affinity purified chicken IgY. Liquid in PBS, pH 7.2 with 0.02% sodium azide.

Storage and Stability

Maintain at 2-8°C in undiluted aliquots for up to 6 months after date of receipt. It is recommended that you store the antibody in the dark.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Caroline Guglielmetti et al.
Proceedings of the National Academy of Sciences of the United States of America, 114(33), E6982-E6991 (2017-08-02)
Proinflammatory mononuclear phagocytes (MPs) play a crucial role in the progression of multiple sclerosis (MS) and other neurodegenerative diseases. Despite advances in neuroimaging, there are currently limited available methods enabling noninvasive detection of MPs in vivo. Interestingly, upon activation and
The 4.1B cytoskeletal protein regulates the domain organization and sheath thickness of myelinated axons.
Einheber, S; Meng, X; Rubin, M; Lam, I; Mohandas, N; An, X; Shrager, P; Kissil, J; Maurel et al.
Glia null
Elodie Martin et al.
Brain plasticity (Amsterdam, Netherlands), 5(2), 123-133 (2020-12-08)
Microglia are the resident macrophages of the central nervous system (CNS). In multiple sclerosis (MS) and related experimental models, microglia have either a pro-inflammatory or a pro-regenerative/pro-remyelinating function. Inhibition of Bruton's tyrosine kinase (BTK), a member of the Tec family
Debbie Le Blon et al.
Journal of neuroinflammation, 13(1), 288-288 (2016-11-11)
Promoting the neuroprotective and repair-inducing effector functions of microglia and macrophages, by means of M2 polarisation or alternative activation, is expected to become a new therapeutic approach for central nervous system (CNS) disorders in which detrimental pro-inflammatory microglia and/or macrophages
Krystyna M Wozniak et al.
Cancer research, 78(3), 817-829 (2017-12-02)
Chemotherapy-induced peripheral neuropathy (CIPN) is a major cause of disability in cancer survivors. CIPN investigations in preclinical model systems have focused on either behaviors or acute changes in nerve conduction velocity (NCV) and amplitude, but greater understanding of the underlying

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service