Skip to Content
Merck
All Photos(1)

Key Documents

AB9210

Sigma-Aldrich

Anti-Insulin Degrading Enzyme Antibody

serum, Chemicon®

Synonym(s):

IDE, Insulysin, Insulinase

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

serum

antibody product type

primary antibodies

clone

polyclonal

species reactivity

rat

packaging

antibody small pack of 25 μL

manufacturer/tradename

Chemicon®

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

rat ... Ins1(24505)

Specificity

Insulin Degrading Enzyme (IDE). It is expected that the antibody will also react with human and mouse due to immunogen sequence homology.

Immunogen

Synthetic peptide from rat IDE.

Application

Anti-Insulin Degrading Enzyme Antibody is an antibody against Insulin Degrading Enzyme for use in WB.
Research Category
Neuroscience
Research Sub Category
Neurodegenerative Diseases
Western blotting: 1:10,000-1:20,000. The antibody recognizes a protein of ~110 kDa on rat liver tissue lysate.

Optimal working dilutions must be determined by the end user.

Quality

Tested

Physical form

Rabbit serum. Liquid.

Storage and Stability

Maintain at -20°C in undiluted aliquots for up to 6 months after date of receipt. Avoid repeated freeze/thaw cycles.

Other Notes

Please refer to lot specific datasheet.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Beatriz Merino et al.
Metabolism: clinical and experimental, 113, 154352-154352 (2020-09-12)
The insulin-degrading enzyme (IDE) is a metalloendopeptidase with a high affinity for insulin. Human genetic polymorphisms in Ide have been linked to increased risk for T2DM. In mice, hepatic Ide ablation causes glucose intolerance and insulin resistance when mice are
Miriam Corraliza-Gómez et al.
Cells, 11(2) (2022-01-22)
The insulin-degrading enzyme (IDE) is a zinc-dependent metalloendopeptidase that belongs to the M16A metalloprotease family. IDE is markedly expressed in the brain, where it is particularly relevant due to its in vitro amyloid beta (Aβ)-degrading activity. The subcellular localization of
Carlos M González-Casimiro et al.
Cells, 10(9) (2021-09-29)
Insulin-degrading enzyme (IDE) is a highly conserved and ubiquitously expressed Zn2+-metallopeptidase that regulates hepatic insulin sensitivity, albeit its regulation in response to the fasting-to-postprandial transition is poorly understood. In this work, we studied the regulation of IDE mRNA and protein
Enrique Jurado-Ruiz et al.
Scientific reports, 9(1), 11311-11311 (2019-08-07)
Dietary composition plays an important role in the pathophysiology of type 2 diabetes. Monounsaturated fatty acid consumption has been positively associated with improved insulin sensitivity and β-cell function. We examined whether an extra virgin olive oil (EVOO) high fat diet
Jéssica Taday et al.
Metabolites, 14(3) (2024-03-27)
Astrocytes play fundamental roles in the maintenance of brain homeostasis. The dysfunction of these cells is widely associated with brain disorders, which are often characterized by variations in the astrocyte protein markers GFAP and S100B, in addition to alterations in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service