Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
UNSPSC Code:
12352203
NACRES:
NA.41
eCl@ss:
32160702
Conjugate:
unconjugated
Clone:
polyclonal
Application:
ICC, IHC, WB
Citations:
10
biological source
rabbit
Quality Level
conjugate
unconjugated
antibody form
affinity purified immunoglobulin
antibody product type
primary antibodies
clone
polyclonal
purified by
affinity chromatography
species reactivity
rat, mouse
manufacturer/tradename
Chemicon®
technique(s)
immunocytochemistry: suitable, immunohistochemistry: suitable, western blot: suitable
NCBI accession no.
UniProt accession no.
shipped in
wet ice
target post-translational modification
unmodified
Gene Information
human ... SCN8A(6334)
General description
Voltage-gated sodium channels perform critical roles for electrical signaling in the nervous ssystem by generating action potentials in axons and dndrites. At least 10 genes encode sodium channels in mammals, but specific physiological roles that distinguish each of these isoforms are not well known. Nav1.6 is highly concentrated at notes of Ranvier of both sensory and motor axons in the peripheral nervous system and at nodes in the central nervous system. Because of its consistant and restrictied labeling, Anti-Nav1.6 (AB5580) has been used effectively as a nodes of Ranvier marker.
Immunogen
Peptide corresponding to amino acids 1042-1061 from mouse (Accession number AAD20438) or rat (Accession number AAC26014) Scn8a.
Application
Detect SCN8A Sodium Channel using this Anti-SCN8A Sodium Channel Antibody validated for use in IC, IH & WB.
Western blot: (1:200) using ECL on rat brain membranes.
Immunohistochemistry on fixed frozen sections.
Immunocytochemistry on rat dorsal root ganglion primary culture (1:100)
Dilutions should be made using a carrier protein such as BSA (1-3%)
Optimal working dilutions must be determined by the end user.
Immunohistochemistry on fixed frozen sections.
Immunocytochemistry on rat dorsal root ganglion primary culture (1:100)
Dilutions should be made using a carrier protein such as BSA (1-3%)
Optimal working dilutions must be determined by the end user.
Biochem/physiol Actions
Recognizes Scn8a (Nav1.6). The epitope corresponds to the intracellular loop between II and III domains of Scn8a. Does not cross react with any other known proteins.
SPECIES REACTIVITIES: It is expected that the antibody will also work on mouse (20/20) and human (19/20) due to sequence homology. Other species have not been tested.
SPECIES REACTIVITIES: It is expected that the antibody will also work on mouse (20/20) and human (19/20) due to sequence homology. Other species have not been tested.
Other Notes
Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.
Legal Information
CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany
Still not finding the right product?
Try our Product Selector Tool to narrow your options
hcodes
pcodes
Hazard Classifications
Aquatic Chronic 3
Storage Class
11 - Combustible Solids
wgk
WGK 3
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Jacy L Wagnon et al.
Neurology. Genetics, 3(4), e170-e170 (2017-07-14)
To determine the functional effect of SCN8A missense mutations in 2 children with intellectual disability and developmental delay but no seizures. Genomic DNA was analyzed by next-generation sequencing. SCN8A variants were introduced into the Nav1.6 complementary DNA by site-directed mutagenesis.
Ping Li et al.
eNeuro, 4(3) (2017-07-01)
The GABA-B receptor is densely expressed throughout the brain and has been implicated in many CNS functions and disorders, including addiction, epilepsy, spasticity, schizophrenia, anxiety, cognitive deficits, and depression, as well as various aspects of nervous system development. How one
Devid Damiani et al.
The Journal of comparative neurology, 520(7), 1406-1423 (2011-11-22)
Retinitis pigmentosa (RP) is a family of inherited diseases causing progressive photoreceptor death. Retinal ganglion cells (RGCs) form the biological substrate for various therapeutic approaches designed to restore vision in RP individuals. Assessment of survival and preservation of RGCs in
Global Trade Item Number
| SKU | GTIN |
|---|---|
| AB5580-200UL | 04053252332821 |
| AB5580-50UL | 04053252476778 |