Skip to Content
Merck
All Photos(1)

Key Documents

AB5122

Sigma-Aldrich

Anti-Adrenergic Receptor Antibody, β 3

Chemicon®, from rabbit

Synonym(s):

Anti-Anti-B3AR, Anti-Anti-GPCR

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human

manufacturer/tradename

Chemicon®

technique(s)

ELISA: suitable
immunohistochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... ADRB3(155)

Specificity

Human beta 3 adrenergic receptor (B3AR) is a member of the G-protein coupled receptor superfamily. They typically contain 7 transmembrane domains (Barak et al., 1995) Human B3AR is 408 aa membrane protein (Emorine et al., 1989; Grannenman et al., 1992; Van Spronsen et al., 1993; Lelias et al., 1993).

Immunogen

A 19 aa sequence at the C-terminus of the human B3AR, splce variant 3 (Emorine et al., 1989; Grannenman et al., 1992; Van Spronsen et al., 1993; Lelias et al., 1993). Control peptide is available under catalog number AG388.

Application

Anti-Adrenergic Receptor Antibody, β 3 detects level of Adrenergic Receptor & has been published & validated for use in ELISA, IH & WB.
Western blot: 1-10 μg/mL using Chemiluminescence technique

ELISA: 1:10,000 - 100,000

Immunohistochemistry: 2-20 μg in formaldehyde fixed tissue.

Optimal working dilutions must be determined by end user.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

C Rouget et al.
The Journal of clinical endocrinology and metabolism, 90(3), 1644-1650 (2004-12-09)
To assess whether pregnancy might influence the functionality and expression of human myometrial beta(2)- and beta(3)-adrenoceptors (beta(2)- and beta(3)-AR), we performed functional, binding, Western blot, and molecular biology experiments in human nonpregnant and near-term pregnant myometrium. Inhibition of spontaneous contractions
Alessandro De Stefano et al.
Vascular pharmacology, 146, 107094-107094 (2022-08-08)
As inadequate perfusion has emerged as a key determinant of adipose tissue dysfunction in obesity, interest has grown regarding possible pharmacological interventions to prevent this process. Mirabegron has proved to improve insulin sensitivity and glucose homeostasis in obese humans via
Ting Sun et al.
Haematologica, 105(3), 661-673 (2019-07-11)
The role of the bone marrow niche in essential thrombocythemia (ET) remains unclear. Here, we observed multilevel defects in the hematopoietic niche of patients with JAK2V617F-positive ET, including functional deficiency in mesenchymal stromal cells (MSC), immune imbalance, and sympathetic-nerve damage.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service