Skip to Content
Merck

AB3100

Anti-Orexin-B Antibody

Chemicon®, from rabbit

Synonym(s):

Anti-NRCLP1, Anti-OX, Anti-PPOX

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
eCl@ss:
32160702
Conjugate:
unconjugated
Clone:
polyclonal
Application:
ELISA, WB
Citations:
1
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

rabbit

Quality Segment

conjugate

unconjugated

antibody form

affinity purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

bovine, mouse, pig, rat, canine

manufacturer/tradename

Chemicon®

technique(s)

ELISA: suitable, western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... PPOX(5498)

General description

Several peptides associated with feeding behavior have been reported recently. Orexins (Orexin-A and Orexin-B) are a family of hypothalamic neuropeptides selectively expressed in the hypothalamus (Sakurai et al., 1998). Orexin-A and Orexin-B are derived from the same precursor (Prepro-orexin) by proteolytic cleavage. Prepro-orexin is an 130 aa peptide with a putative 33 aa secretory sequence, a hydrophobic core followed by residues with small polar side chains. The expression was detected in brain and to a small extent in testis (Sakurai et al., 1998; DeLecca et al., 1998). These neuropeptides bind and activate two closely related Orexin receptors--G protein coupled receptors (GPCRs) OX1R and OX2R.

Immunogen

A 13 amino acid peptide mapping near the amino terminus of mouse Orexin-B. The immunogen peptide has 92% similarity with human Orexin-B.

Application

Anti-Orexin-B Antibody detects level of Orexin-B & has been published & validated for use in ELISA & WB.
Western blot: 1-10 μg/mL using Chemiluminescence technique

ELISA: 1:10,000 - 100,000 using 50-100 ng of control peptide per well.

Optimal working dilutions must be determined by end user.

Biochem/physiol Actions

Mouse/Rat Orexin-B. Reacts with full length Orexin-B. The AB3100 epitope is not found in Orexin-A. Since Orexins-A and Orexin-B are derived from an unprocessed pre-pro orexins (130 aa), the anti-Orexin-B antibodies may also react with pre-pro orexins.

Physical form

Affinity purified immunoglobulin. Liquid in PBS, with 0.1% BSA .

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany


Still not finding the right product?

Explore all of our products under Anti-Orexin-B Antibody

or

Try our Product Selector Tool to narrow your options


Storage Class

12 - Non Combustible Liquids

wgk

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library