Skip to Content
Merck

AB3085

Anti-Na+/H+ Exchanger-3 Antibody

Chemicon®, from rabbit

Synonym(s):

NHE3

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
eCl@ss:
32160702
Conjugate:
unconjugated
Clone:
polyclonal
Application:
ELISA, WB
Citations:
16
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

rabbit

Quality Segment

conjugate

unconjugated

antibody form

affinity purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

rat

manufacturer/tradename

Chemicon®

technique(s)

ELISA: suitable, western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

rat ... Slc9A3(24784)

General description

93 kDa

Immunogen

A 22 AA peptide within the cytoplasmic, c-terminal domain of the rat NHE3 (Brant et al. 1995; Orlowsji et al. 1992; Chris Yun et al. 1995; Josette & Pouyssegur 1995). Control peptide is available, cat# AG779.

Application

Research Category
Neuroscience
Research Sub Category
Ion Channels & Transporters
This Anti-Na+/H+ Exchanger-3 Antibody is validated for use in ELISA, WB for the detection of Na+/H+ Exchanger-3.
Western blot: 1-10 μg/mL using Chemiluminescence technique. A band of approximately 90 kD has been detected in brush border membrane (Yohsioka et al. 1997). ELISA: 1:100,000 using 50-100 ng control peptide/well.

Optimal working dilutions must be determined by end user.

Biochem/physiol Actions

Recognizes NHE3.
The immunogen sequence shares 90% homology with mouse NHE3.

Physical form

Affinity Purified immunoglobulin. Liquid in PBS with 0.1% BSA.
ImmunoAffinity Purified

Preparation Note

Maintain for 6 months at -20°C from date of shipment. Aliquot to avoid repeated freezing and thawing. For maximum recovery of product, centrifuge the original vial after thawing and prior to removing the cap.

Analysis Note

Control
Kidney, jejunum, colon, and stomach tissue

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.


Still not finding the right product?

Try our Product Selector Tool to narrow your options


Storage Class

12 - Non Combustible Liquids

wgk

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library