Skip to Content
Merck
All Photos(4)

Key Documents

AB3080

Sigma-Aldrich

Anti-Green Fluorescent Protein Antibody

Chemicon®, from rabbit

Synonym(s):

GFP, eGFP

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

affinity purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

species reactivity

vertebrates

manufacturer/tradename

Chemicon®

technique(s)

ELISA: suitable
immunocytochemistry: suitable
immunohistochemistry: suitable
western blot: suitable

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

General description

The Green Fluorescent Protein (GFP) from the jellyfish Aequorea victoria is used as a fluorescent indicator for monitoring gene expression in a variety of cellular systems, including living organisms and fixed tissues. Unlike other bioluminescent reporters, GFP fluoresces in the absence of substrates, cofactors, or other intrinsic or extrinsic proteins. Purified GFP is a 27 kDa monomer consisting of 238 amino acids and emits green light (emission maximum at 509 nm) when excited with blue or UV light.

Specificity

AB3080 is made against highly-purified native GFP from Aequorea victoria. It is reactive with GFP from both native and recombinant sources. While other commercial antibodies to GFP typically demonstrate strong reactivity with E. coli, the AB3080 GFP antibody has been purified by Protein A chromatography, followed by extensive adsorption against whole E. coli BL21 strain cells, giving them minimal cross-reactivity with E. coli.

Immunogen

Highly purified native GFP from Aequorea victoria.

Application

Research Category
Epitope Tags & General Use
Research Sub Category
Epitope Tags
This Anti-Green Fluorescent Protein Antibody is validated for use in ELISA, IC, IH, WB for the detection of Green Fluorescent Protein.
Western Blot:
A previous lot of this antibody was used in western blot.

ELISA:
A 1:2000-1:10,000 dilution of a previous lot was used in Enzyme-linked Immunosorbent Assay.

Note: While AB3080 has been adsorbed against E. coli proteins, most secondary antibodies have not. To reduce the likelihood of E. coli reactivity from a secondary antibody, we suggest the use of Millipore’s highly-purified secondary antibodies in indirect detection systems.

Immunocytochemistry:

Optimal working dilutions must be determined by end user.

Quality

Routinely evaluated by immunocytochemistry on RENCX cells.

Immunocytochemistry Analysis: Confocal fluorescent analysis of RENCX constitutively expressed GFP using this lot of anti-GFP Rabbit pAB (Red). Green indicates the expressed GFP.

Target description

GFP is 27 kDa Dependent upon the molecular weight of the fusion protein being detected.

Physical form

Format: Purified
Protein A purified
Purified rabbit polyclonal IgG in 10 mM Tris buffer, pH 7.2 with 0.01% sodium azide.

Storage and Stability

Stable for 1 year at -20ºC from date of receipt.

Handling Recommendations:
Upon receipt, and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.

Analysis Note

Control
GFP fusion protein expressed in cells

RENCX cells

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Subha Karumuthil-Melethil et al.
Human gene therapy, 27(7), 509-521 (2016-05-21)
GM2 gangliosidosis is a family of three genetic neurodegenerative disorders caused by the accumulation of GM2 ganglioside (GM2) in neuronal tissue. Two of these are due to the deficiency of the heterodimeric (α-β), "A" isoenzyme of lysosomal β-hexosaminidase (HexA). Mutations
Noninvasive in vivo imaging of monocyte trafficking to atherosclerotic lesions.
Kircher, MF; Grimm, J; Swirski, FK; Libby, P; Gerszten, RE; Allport, JR; Weissleder, R
Circulation null
Aosa Kamezaki et al.
Scientific reports, 6, 28873-28873 (2016-07-02)
Neuregulin1 (NRG1) plays diverse developmental roles and is likely involved in several neurological disorders including schizophrenia. The transmembrane NRG1 protein is proteolytically cleaved and released as a soluble ligand for ErbB receptors. Such post-translational processing, referred to as 'ectodomain shedding'
Targeted ablation and reorganization of the principal preplate neurons and their neuroblasts identified by golli promoter transgene expression in the neocortex of mice.
Xie, YY; Jacobs, E; Fisher, R
ASN Neuro null
Induction of spine growth and synapse formation by regulation of the spine actin cytoskeleton.
Zito, K; Knott, G; Shepherd, GM; Shenolikar, S; Svoboda, K
Neuron null

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service