Skip to Content
Merck
All Photos(1)

Key Documents

AB2201

Sigma-Aldrich

Anti-mPER1 (residues 6-21) Antibody

serum, from rabbit

Synonym(s):

Circadian clock protein PERIOD 1, Circadian pacemaker protein Rigui, Period, drosophila, homolog of period 1, period (Drosophila) homolog 1, period homolog 1 (Drosophila)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

serum

antibody product type

primary antibodies

clone

polyclonal

species reactivity

human, mouse, rat

technique(s)

western blot: suitable

GenBank accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

mouse ... Per1(18626)

General description

This gene is a member of the Period family of genes and is expressed in a circadian pattern in the suprachiasmatic nucleus, the primary circadian pacemaker in the mammalian brain. Genes in this family encode components of the circadian rhythms of locomotor activity, metabolism, and behavior. Circadian expression in the suprachiasmatic nucleus continues in constant darkness, and a shift in the light/dark cycle evokes a proportional shift of gene expression in the suprachiasmatic nucleus. The specific function of this gene is not yet known. Alternative splicing has been observed in this gene; however, these variants have not been fully described.

Specificity

Cat. # AB2201 will recognize the N-terminus of mPER1.
May react with Drosophila, Arvicanthis niloticus and Mesocricetus auratus based on sequence homology. Reactivity with other species has not been tested.

Application

Detect mPER1 (residues 6-21) using this Anti-mPER1 (residues 6-21) Antibody validated for use in WB.
Immunohistichemistry:
Tested on whole brain (SCN; peak staining at CT/ZT12). Optimium working dilutions need to be determined by end used.

Quality

Routinely tested by western blot on human brain lysate

Target description

136 kDa

Legal Information

GenBank is a registered trademark of United States Department of Health and Human Services

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nobuko Katoku-Kikyo et al.
PLoS biology, 21(12), e3002419-e3002419 (2023-12-04)
Circadian regulation of gene expression is prevalent and plays critical roles in cell differentiation. However, its roles in the reprogramming of differentiated cells remain largely unknown. Here, we found that one of the master circadian regulators PER1 promoted virus-mediated reprogramming
Wenhao Ge et al.
Journal of lipid research, 64(6), 100390-100390 (2023-05-21)
Several epidemiological studies suggest a correlation between eating time and obesity. Night eating syndrome characterized by a time-delayed eating pattern is positively associated with obesity in humans as well as in experimental animals. Here, we show that oil intake at
Yasuhiro Umemura et al.
Proceedings of the National Academy of Sciences of the United States of America, 119(1) (2021-12-22)
In mammals, circadian clocks are strictly suppressed during early embryonic stages, as well as in pluripotent stem cells, by the lack of CLOCK/BMAL1-mediated circadian feedback loops. During ontogenesis, the innate circadian clocks emerge gradually at a late developmental stage, and
Tomoko Ikeno et al.
Journal of biological rhythms, 31(4), 352-364 (2016-04-15)
In mammals, the circadian system is composed of a principal circadian oscillator located in the suprachiasmatic nucleus (SCN) and a number of subordinate oscillators in extra-SCN brain regions and peripheral tissues/organs. However, how the time-keeping functions of this multiple oscillator
Floriana Mogavero et al.
The European journal of neuroscience, 55(6), 1492-1503 (2022-03-02)
Although aggression has been linked to disturbances of circadian rhythm, insight into the neural substrate of this association is currently lacking. The suprachiasmatic nucleus (SCN) of the hypothalamus, the master circadian clock, is regulated by clock genes and known to

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service