Skip to Content
Merck

AB1920

Anti-Integrin α3 Antibody

serum, Chemicon®

Synonym(s):

CD49c

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
eCl@ss:
32160702
Conjugate:
unconjugated
Clone:
polyclonal
Application:
ELISA, IF, IHC, IP, RIA, WB
Citations:
86
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

rabbit

Quality Segment

conjugate

unconjugated

antibody form

serum

antibody product type

primary antibodies

clone

polyclonal

species reactivity

porcine, rat, goat, canine, sheep, hamster, equine, pig, bovine, human, mouse, chicken, horse

manufacturer/tradename

Chemicon®

technique(s)

ELISA: suitable, immunofluorescence: suitable, immunohistochemistry: suitable, immunoprecipitation (IP): suitable, radioimmunoassay: suitable, western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... ITGA3(3675)

General description

130 kDa
Integrins are a family of dimeric, transmembrane proteins that mediate cell-cell and extracellular matrix adhesion. Signals transduced by integrins play a role in many biological processes, including cell growth, differentiation, migration and apoptosis. The integrin family is composed of at least 19 alpha and 8 beta subunits that form over twenty different alpha-beta non-covalently bound dimeric combinations on the cell surface. Both of the subunits contribute to the binding of the ligand. The alpha-3 subunit (CD49c) is highly concentrated in epithelial cells where it strongly adheres to Laminin-5 and Laminin-5 induced rapid adhesion can be blocked by antibodies against the alpha-3 integrin subunit. The alpha-3 subunit exists in two different splice variants, denoted as "A" and "B". The only difference that results from this differential splicing is a total change in the cytoplasmic domain, while the extracellular domain stays the same. Knock-out mice lacking this subunit show prenatal lethality and abnormalities in the kidneys.

Immunogen

A synthetic peptide (CRIQPSETERLTDDY) derived from the COOH terminal sequence located in the cytoplasmic domain of the human alpha3 integrin subunit.

Application

Detect Integrin α3 using this Anti-Integrin α3 Antibody validated for use in ELISA, IF, IH, IP, RIA & WB.
ELISA/RIA:
A 1:1,000 dilution of a previous lot was used in Enzyme-linked Immunosorbent Assay/Radioimmunoassay.

Immunoprecipitation:
5 µL of a previous lot precipitated alpha3/beta1 from 5 x 106 cells (Sherry, D.M., 2001).

Immunohistochemistry/immunofluorescence:
1:500 dilution of a previous lot was used, suggest use on acetone-fixed tissue.

Western Blot Analysis:
1:500 dilution of a previous lot was used.

Optimal working dilutions must be determined by end user.

Biochem/physiol Actions

Integrin alpha3. Specificity determined by immunoprecipitation from a detergent extract of 35S-methionine labeled neuroblastoma cells. No cross-reactivity to alpha1, alpha2, alpha4, alpha5, alpha6, or alphaV integrins.
Reactivity with other species has not been determined.

Physical form

Rabbit polyclonal IgG antiserum containing 0.05% sodium azide.

Analysis Note

Control
Human fetal foreskin fibroblasts (HFFF), neuroblastomas
Routinely evaluated by Western Blot on A431 lysates.

Western blot: 1:500 dilution of this lot detected INTEGRIN ALPHA 3 on 10 μg of A431 lysates.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.
Replaces: AB1948

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany


Still not finding the right product?

Try our Product Selector Tool to narrow your options


Storage Class

10 - Combustible liquids

wgk

WGK 1



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library