Skip to Content
Merck
All Photos(1)

Key Documents

AB19015

Sigma-Aldrich

Anti-MMP-2 Antibody, catalytic domain

Chemicon®, from rabbit

Synonym(s):

Gelatinase A, 72 kDa Type IV Collagenase

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

species reactivity

human, mouse, rat

manufacturer/tradename

Chemicon®

technique(s)

immunoprecipitation (IP): suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... MMP2(4313)

Specificity

Recognizes rat MMP-2. Exhibits no cross-reactivity with other MMP family members. The antibody was generated using E. coli-expressed active rat 72 kDa type IV collagenase (catalytic domain) as an immunogen.

Immunogen

E. coli-expressed active rat 72 kDa type IV collagenase (catalytic domain)
Epitope: catalytic domain

Application

Detect MMP-2 using this Anti-MMP-2 Antibody, catalytic domain validated for use in IP & WB.
Research Category
Cell Structure
Research Sub Category
MMPs & TIMPs
Western blot: 1:2,000

Immunoprecipitation: 1:500

MMP-2 [Gelatinase-A] is constitutively produced in quiescent cells and tissues, and the enzyme has a high specific activity against denatured collagen. The low protein levels produced in cell culture (pg/mL) are often below the threshold of detection by standard Western Blotting. The enzyme can be concentrated from culture media by gelatin-agarose affinity chromatography (Goldberg, 1992) or immunoprecipitation. This can prevent appearance of a spurious band of antibody binding in the vicinity of 68 kDa displayed by some concentrated media.

Optimal working dilutions must be determined by end user.

Linkage

Replaces: 04-1048

Physical form

Format: Purified
Purified immunoglobulin. Liquid in PBS, containing 0.05% sodium azide.

Storage and Stability

Maintain refrigerated at 2-8°C in undiluted aliquots for up to 12 months.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jennifer A Kwan et al.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology, 18(6), 690-692 (2004-02-10)
Matrix metalloproteinases (MMPs) are traditionally known for their role in extracellular matrix remodeling. Increasing evidence reveals several alternative substrates and novel biological roles for these proteases. Recent evidence showed the intracellular localization of MMP-2 within cardiac myocytes, colocalized with troponin
John R Barbour et al.
The Journal of thoracic and cardiovascular surgery, 132(4), 788-795 (2006-09-27)
An important component of matrix remodeling during thoracic aortic aneurysm progression is the balance between matrix metalloproteinases and their endogenous inhibitors (tissue inhibitors of metalloproteinases). However, whether and to what degree matrix metalloproteinase/tissue inhibitor of metalloproteinases profiles change over time
Vascular cell adhesion molecule 1 (VCAM-1) activation of endothelial cell matrix metalloproteinases: role of reactive oxygen species.
Deem, TL; Cook-Mills, JM
Blood null
Yu-Mei Xue et al.
Molecular medicine reports, 17(2), 3425-3431 (2017-12-20)
Atrial fibrosis is the fundamental characteristic of the structural pathology associated with atrial fibrillation (AF). Inflammation can contribute to atrial fibrosis, engendering AF. The present study aimed to investigate the role of macrophage migration inhibitory factor (MIF), a pleiotropic cytokine
Hsien-Feng Liao et al.
PloS one, 12(9), e0185021-e0185021 (2017-09-26)
Chemotherapy insensitivity continues to pose significant challenges for treating non-small cell lung cancer (NSCLC). The purposes of this study were to investigate whether 3,6-dimethoxy-1,4,5,8-phenanthrenetetraone (NCKU-21) has potential activity to induce effective toxicological effects in different ethnic NSCLC cell lines, A549

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service