Skip to Content
Merck

AB1871

Anti-Caspase 1 Antibody

Chemicon®, from rabbit

Synonym(s):

ICE

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
eCl@ss:
32160702
Conjugate:
unconjugated
Clone:
polyclonal
Application:
IF, IP, WB
Citations:
29
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

rabbit

conjugate

unconjugated

antibody form

affinity purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human, rat, mouse

manufacturer/tradename

Chemicon®

technique(s)

immunofluorescence: suitable, immunoprecipitation (IP): suitable, western blot: suitable

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... CASP1(834)
mouse ... Casp1(12362)
rat ... Casp1(25166)

General description

Caspase-1 (ICE, interleukin-1b-converting enzyme) is the mammalian caspase responsible for the proteolytic conversion of the proforms of interleukin-1b and IL-18 into active cytokines. Like other caspases, caspase-1 itself is synthesized as a proenzyme that is cleaved during activation into large (20 kDa) and small (10 kDa) subunits.

Application

Western blot: 1-4 μg/mL

Immunoprecipitation: 4-8 μg/mL

Immunofluorescence: 10-20 μg/mL

Optimal working dilutions must be determined by end user.
This Anti-Caspase 1 Antibody is validated for use in IF, IP, WB for the detection of Caspase 1.

Biochem/physiol Actions

AB1871 recognizes the 45 kDa procaspase-1 as well as the 20 kDa cleaved caspase-1.

Physical form

Affinity purified immunoglobulin. Liquid in PBS, pH 7.2, containing 50% glycerol, 1% BSA and 0.02% sodium azide.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany


Still not finding the right product?

Explore all of our products under Anti-Caspase 1 Antibody

or

Try our Product Selector Tool to narrow your options


Storage Class

10 - Combustible liquids

wgk

WGK 2



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library