Skip to Content
Merck
All Photos(1)

Key Documents

AB1840

Sigma-Aldrich

Anti-Interleukin-8 Antibody

serum, Chemicon®

Synonym(s):

IL-8

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

serum

antibody product type

primary antibodies

clone

polyclonal

species reactivity

sheep

manufacturer/tradename

Chemicon®

technique(s)

ELISA: suitable

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

sheep ... Cxcl8(443418)

Specificity

Reacts with ovine IL-8. No reactivity detected to recombinant ovine IL-1beta, IL-6, MCP or TNFalpha.

Immunogen

Recombinant ovine IL-8

Application

Anti-Interleukin-8 Antibody detects level of Interleukin-8 & has been published & validated for use in ELISA.
Detecting antibody in ELISA, using monoclonal anti-ovine IL-8 as capture antibody. Recommended dilution factor 1:500. See suggested protocol.

Optimal working dilutions must be determined by the end user.

Suggested ELISA Protocol For Ovine IL-8

1. Coat high capacity microtiter wells (e.g. Nunc maxisorp) with monoclonal anti-IL-8 antibody (Chemicon Catalog Number MAB1044) at a concentration of 5 μg/mL in 0.1 M carbonate buffer pH 9.6 overnight at 4ºC.

2. Wash wells 3 x with PBS + 0.05% Tween 20 (PBST).

3. Add samples to wells. Incubate at room temperature in a humidified box for 1 hour. Wash 3 x with PBST.

4. Dilute rabbit anti-IL-8 (Chemicon Catalog Number AB1840) 1:500 in PBST and add 100 μL to wells. Incubate at room temperature for 30 minutes. Wash 3 x with PBST.

5. Dilute anti-rabbit IgG-HRP to appropriate level in PBST and add 100 μL to wells. Incubate 30 minutes at room temperature. Wash 3 x with PBST.

6. Add 100 μL of TMB chromagen to each well. Incubate for 5-10 minutes and stop reaction with 50 μL of 2M sulfuric acid.

Notes:

The anti-rabbit IgG HRP must be adsorbed against mouse immunoglobulin, and should be checked for non-reactivity with coated plates.

Non-specific binding may be reduced by addition of protein such as skim milk powder to the diluents.

Assay sensitivity may be altered by adjusting incubation times and reagent concentrations.
Research Category
Inflammation & Immunology
Research Sub Category
Cytokines & Cytokine Receptors

Physical form

Rabbit serum. Liquid containing 0.1% sodium azide.

Storage and Stability

Maintain at -20°C in undiluted aliquots for up to 12 months. Avoid repeated freeze/thaw cycles.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yong Zhu et al.
Biology of reproduction, 104(2), 317-324 (2020-12-11)
High unintended pregnancy rates are partially due to lack of effective nonhormonal contraceptives; development of safe, effective topical vaginal methods will address this need. Preclinical product safety and efficacy assessment requires in vivo testing in appropriate models. The sheep is
Selective exposure of the fetal lung and skin/amnion (but not gastro-intestinal tract) to LPS elicits acute systemic inflammation in fetal sheep.
Kemp, MW; Kannan, PS; Saito, M; Newnham, JP; Cox, T; Jobe, AH; Kramer, BW; Kallapur, SG
Testing null
Tate Gisslen et al.
Innate immunity, 20(2), 214-224 (2013-06-12)
Histologic chorioamnionitis, frequently associated with preterm births and adverse outcomes, results in prolonged exposure of preterm fetuses to infectious agents and pro-inflammatory mediators, such as LPS. Endotoxin tolerance-type effects were demonstrated in fetal sheep following repetitive systemic or intra-amniotic (i.a.)
Clare A Berry et al.
Reproductive sciences (Thousand Oaks, Calif.), 18(11), 1092-1102 (2011-04-16)
We tested the hypothesis that interleukin 1 (IL-1) mediates intra-amniotic lipopolysaccharide (LPS)-induced chorioamnionitis in preterm fetal sheep. Time-mated Merino ewes with singleton fetuses received IL-1α, LPS, or saline (control) by intra-amniotic injection 1 to 2 days before operative delivery at
Monique G M Willems et al.
American journal of physiology. Lung cellular and molecular physiology, 310(1), L1-L7 (2015-11-01)
Chorioamnionitis, an inflammatory reaction of the fetal membranes to microbes, is an important cause of preterm birth and associated with inflammation-driven lung injury. However, inflammation in utero overcomes immaturity of the premature lung by inducing surfactant lipids and lung gas

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service