Skip to Content
Merck
All Photos(1)

Key Documents

AB1797P

Sigma-Aldrich

Anti-Arginine Vasopressin Receptor V2 Antibody

Chemicon®, from rabbit

Synonym(s):

AVPR-V2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

affinity purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

rat

manufacturer/tradename

Chemicon®

technique(s)

ELISA: suitable
immunohistochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... AVPR2(554)

Specificity

Arginine vasopressin receptor (AVP-V2), rat. The peptide used for immunization has no significant similarity with AVP V1a or V1b receptors.

Immunogen

21 amino acid peptide from the 3rd cytoplasmic domain of rat AVP-V2 (Lolait et al. 1992; Fisov et al. 1994).

Application

Anti-Arginine Vasopressin Receptor V2 Antibody is an antibody against Arginine Vasopressin Receptor V2 for use in ELISA, IH & WB.
Research Category
Neuroscience
Research Sub Category
CNS Control of Metabolism

Hormones & Receptors
Western blot: 1-10 μg/mL using ECL. An antibody raised against this peptide has detected a glycosylated band at ~62 kDa in membranes from rat inner medulla (Nonoguchi et al. 1995). N-Glycanase reduced the size to ~36 kDa (deglycosylated form).

Immunohistochemistry: 2-10 μg/mL on paraformaldehyde fixed tissue. AVP-V2 has been localized in terminal IMCD and other areas of rat kidney (Nonoguchi et al. 1995).

ELISA: 0.5-1.0 μg/mL using 1 μg/mL of control peptide to coat plate.

Optimal working dilutions must be determined by the end user.

Linkage

Replaces: AB3512P

Physical form

Affinity purified immunoglobulin. Liquid in PBS containing 0.1% BSA.

Storage and Stability

Maintain at -20°C in undiluted aliquots for up to six months. Avoid repeated freeze/thaw cycles.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Itaru Goto et al.
Circulation journal : official journal of the Japanese Circulation Society, 80(4), 989-997 (2016-03-01)
The expression of vasopressin type 2 receptor (V2R) in the lung, and the long-term effects of tolvaptan, a selective V2R antagonist, on pulmonary circulation and right ventricular (RV) remodeling in a pulmonary arterial hypertension (PAH) rat model were evaluated. Six-week-old
Harini Ramalingam et al.
Cell metabolism, 33(6), 1234-1247 (2021-04-15)
Autosomal dominant polycystic kidney disease (ADPKD) is a common monogenic disorder marked by numerous progressively enlarging kidney cysts. Mettl3, a methyltransferase that catalyzes the abundant N6-methyladenosine (m6A) RNA modification, is implicated in development, but its role in most diseases is

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service