Skip to Content
Merck
All Photos(1)

Documents

AB1620

Sigma-Aldrich

Anti-Vimentin Antibody

serum, Chemicon®

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

goat

Quality Level

antibody form

serum

antibody product type

primary antibodies

clone

polyclonal

species reactivity

human

species reactivity (predicted by homology)

mammals

manufacturer/tradename

Chemicon®

technique(s)

immunocytochemistry: suitable
immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... VIM(7431)

Specificity

Recognizes vimentin in cultured mammalian cells and tissue sections. AB1620 specifically stains the vimentin band (58 kD) using total cell or tissue extracts in immunoblotting. No reactivity to desmin, keratin, neurofilaments or glial filaments. Shows wide cross-reactivity among mammalian species using various immunolabeling techniques.

Immunogen

Vimentin purified from cultured human foreskin fibroblasts.

Application

Anti-Vimentin Antibody detects level of Vimentin & has been published & validated for use in IC, IH, IH(P) & WB.
Immunohistochemistry: 1:20-1:40. Shows reactivity on frozen tissue sections as well as formalin-fixed paraffin embedded tissue by immunoperoxidase and immunofluorescent assays.

Immunocytochemistry

Western blot

Optimal working dilutions must be determined by end user.
Research Category
Cell Structure
Research Sub Category
Cytoskeleton

Target description

58 kDa

Physical form

Neat goat Serum. Liquid containing 0.1% sodium azide.
Unpurified

Storage and Stability

Maintain for 1 year at -20°C from date of shipment. Aliquot to avoid repeated freezing and thawing. For maximum recovery of product, centrifuge the original vial after thawing and prior to removing the cap.

Analysis Note

Control
Breast carcinoma tissue, Colon, Prostate, Tonsil, Tonsilar lymphoma, Uterus, Ureter

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Soumya Navneet et al.
Redox biology, 24, 101199-101199 (2019-04-27)
Hyperhomocysteinemia (Hhcy), or increased levels of the excitatory amino acid homocysteine (Hcy), is implicated in glaucoma, a disease characterized by increased oxidative stress and loss of retinal ganglion cells (RGCs). Whether Hhcy is causative or merely a biomarker for RGC
Karen A Sap et al.
Molecular & cellular proteomics : MCP, 18(9), 1705-1720 (2019-05-30)
Huntington's disease is caused by a polyglutamine repeat expansion in the huntingtin protein which affects the function and folding of the protein, and results in intracellular protein aggregates. Here, we examined whether this mutation leads to altered ubiquitination of huntingtin
DNA-PK target identification reveals novel links between DNA repair signaling and cytoskeletal regulation.
Kotula, E; Faigle, W; Berthault, N; Dingli, F; Loew, D; Sun, JS; Dutreix, M; Quanz, M
Testing null
Accessing key steps of human tumor progression in vivo by using an avian embryo model.
Hagedorn, M; Javerzat, S; Gilges, D; Meyre, A; de Lafarge, B; Eichmann, A; Bikfalvi, A
Proceedings of the National Academy of Sciences of the USA null
Maribella Domenech et al.
Integrative biology : quantitative biosciences from nano to macro, 4(2), 142-152 (2012-01-12)
Despite strong evidence for the involvement of the stroma in Hedgehog signaling, little is known about the identity of the stromal cells and the signaling mechanisms that mediate the growth promoting effect of Hh signaling. We developed an in vitro

Protocols

Step-by-step culture protocols for neural stem cell culture including NSC isolation, expansion, differentiation and characterization.

Step-by-step culture protocols for neural stem cell culture including NSC isolation, expansion, differentiation and characterization.

Step-by-step culture protocols for neural stem cell culture including NSC isolation, expansion, differentiation and characterization.

Step-by-step culture protocols for neural stem cell culture including NSC isolation, expansion, differentiation and characterization.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service