Skip to Content
Merck
All Photos(1)

Key Documents

AB1569

Sigma-Aldrich

Anti-Dopa Decarboxylase Antibody

Chemicon®, from rabbit

Synonym(s):

Anti-AADC

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

affinity purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

canine, bovine, rat, guinea pig, human, sheep

manufacturer/tradename

Chemicon®

technique(s)

immunohistochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... CDNF(441549)

Specificity

DOPA Decarboxylase (DDC). Reacts with a single band of ~55 kDa by Western blot.

Immunogen

Recombinant bovine DDC expressed in E. coli and purified from inclusion bodies.

Application

Detect Dopa Decarboxylase using this Anti-Dopa Decarboxylase Antibody validated for use in IH & WB.
Immunoblotting: 1:500-1:1,000

Immunohistochemistry using formaldehyde-fixed brain sections.

Optimal working dilutions must be determined by the end user.
Research Category
Neuroscience
Research Sub Category
Neurotransmitters & Receptors

Neuronal & Glial Markers

Physical form

Affinity purified immunoglobulin. Liquid in 10 mM HEPES, pH 7.5, 150 mM NaCl, 100 μg/mL BSA and 50% glycerol.

Storage and Stability

Maintain at -20 to -80°C in undiluted aliquots for up to six months after date of receipt. Store in a NON-self-defrosting freezer. Avoid repeated freeze-thaw cycles.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Branden J Stansley et al.
Neuropharmacology, 67, 243-251 (2012-12-01)
L-dopa is a precursor for dopamine synthesis and a mainstay treatment for Parkinson's disease. However, l-dopa therapy is not without side effects that may be attributed to non-dopaminergic mechanisms. Synthesized dopamine can be neurotoxic through its enzymatic degradation by monoamine
Diana Marie Battaglia et al.
Frontiers in microbiology, 13, 859866-859866 (2022-04-09)
Herpes simplex virus-associated diseases are a complex interaction between cytolytic viral replication and inflammation. Within the normally avascular and immunoprivileged cornea, HSV ocular infection can result in vision-threatening immune-mediated herpetic keratitis, the leading infectious cause of corneal blindness in the
Mengliang Zhang
Neural regeneration research, 11(12), 1904-1909 (2017-02-16)
Monoamine neurotransmitters play an important role in the modulation of sensory, motor and autonomic functions in the spinal cord. Although traditionally it is believed that in mammalian spinal cord, monoamine neurotransmitters mainly originate from the brain, accumulating evidence indicates that
María A Gómez-Muñoz et al.
International journal of molecular sciences, 24(2) (2023-01-22)
Neuroblastoma is a neural crest cell-derived pediatric tumor characterized by high inter- and intra-tumor heterogeneity, and by a poor outcome in advanced stages. Patient-derived xenografts (PDXs) have been shown to be useful models for preserving and expanding original patient biopsies
Kara Kaplan et al.
Journal of applied physiology (Bethesda, Md. : 1985), 120(9), 1070-1081 (2016-02-13)
Genetic deletion of brain serotonin (5-HT) neurons in mice leads to ventilatory deficits and increased neonatal mortality during development. However, it is unclear if the loss of the 5-HT neurons or the loss of the neurochemical 5-HT led to the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service