Skip to Content
Merck
All Photos(1)

Key Documents

AB1526SP

Sigma-Aldrich

Anti-Nerve Growth Factor-β Antibody

Chemicon®, from rabbit

Synonym(s):

NGFbeta

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

species reactivity

chicken, mouse, human, rat

should not react with

bovine

manufacturer/tradename

Chemicon®

technique(s)

ELISA: suitable
immunohistochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... NGF(4803)

Specificity

Nerve growth factor-beta (NGF). Less than 1% cross-reactivity with recombinant human NT3, NT4 and BDNF by ELISA.

Immunogen

Mouse nerve growth factor-beta purified from salivary glands.

Application

Anti-Nerve Growth Factor-β Antibody detects level of Nerve Growth Factor-β & has been published & validated for use in ELISA, WB, IH.
Immunohistochemistry: 1-2 μg/mL (see suggested protocol).

Immunoblotting: 1-2 μg/mL

Inhibition of biological activity: 1-2 μg/mL
BIOLOGICAL ACTIVITY: This antibody will completely inhibit the survival and neurite outgrowth actions of mouse NGF in chicken dorsal root ganglion neurons in vitro.

ELISA: 1-2 μg/mL

Use neat for in vivo animal studies.

Optimal working dilutions must be determined by the end user.
Research Category
Neuroscience
Research Sub Category
Neurochemistry & Neurotrophins

Linkage

Replaces: 04-1119

Physical form

Format: Purified
Purified immunoglobulin. Lyophilized from PBS. Contains no preservative. Reconstitute with 500 μL of sterile distilled water (1 mg/mL).

Storage and Stability

Maintain at -20 to -70°C in undiluted aliquots for up to one year after date of receipt. After reconstitution maintain at -20°C in undiluted aliquots for up to six months. Avoid repeated freeze/thaw cycles. Glycerol (1:1) may be added for additional stability. Avoid repeated freeze/thaw cycles.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Zhi-Hong Wen et al.
European journal of pain (London, England), 24(1), 209-222 (2019-09-09)
Osteoarthritis (OA) is the most common joint disease, especially affecting the knee joint. Etoricoxib, a highly selective cyclooxygenase (COX)-2 inhibitor which can reduce postoperative pain after orthopaedic surgery. The aim of this study was to investigate the effects of oral
Mariana Pehar et al.
The European journal of neuroscience, 24(6), 1575-1580 (2006-09-29)
The p75 neurotrophin receptor (p75NTR) is expressed by degenerating spinal motor neurons in amyotrophic lateral sclerosis (ALS). The mature and pro-form of nerve growth factor (NGF) activate p75NTR to trigger motor neuron apoptosis. However, attempts to modulate p75NTR-mediated neuronal death
D E Lorke et al.
International journal of immunopathology and pharmacology, 23(1), 13-23 (2010-04-10)
To elucidate factors underlying the increased risk of developing Alzheimers disease (AD) in older individuals, the prefrontal cortices of younger (58-79 years) and of older (over 80 years) AD patients were examined by silver impregnation, TUNEL assay and immunohistochemistry for
Ming-Shian Tsai et al.
Experimental & molecular medicine, 50(1), e426-e426 (2018-01-13)
This study investigated the regulatory role of nerve growth factor (NGF) in sirtuin 1 (SIRT1) expression in cholestatic livers. We evaluated the expression of NGF and its cognate receptors in human livers with hepatolithiasis and the effects of NGF therapy
Mariana Pehar et al.
Journal of neurochemistry, 89(2), 464-473 (2004-04-02)
Reactive astrocytes frequently surround degenerating motor neurons in patients and transgenic animal models of amyotrophic lateral sclerosis (ALS). We report here that reactive astrocytes in the ventral spinal cord of transgenic ALS-mutant G93A superoxide dismutase (SOD) mice expressed nerve growth

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service