Skip to Content
Merck

8.55002

Wang resin (100-200 mesh)

Novabiochem®

Synonym(s):

Wang resin (100-200 mesh), p-Benzyloxybenzyl alcohol resin (100-200 mesh), HMP resin (100-200 mesh)

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

CAS Number:
MDL number:
NACRES:
NA.22
UNSPSC Code:
13111023
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Level

product line

Novabiochem®

form

beads

reaction suitability

reaction type: Fmoc solid-phase peptide synthesis

manufacturer/tradename

Novabiochem®

application(s)

peptide synthesis

functional group

alcohol

storage temp.

2-30°C

General description

Standard resin for the batch-wise synthesis of peptide acids by Fmoc SPPS.

Associated Protocols and Technical Articles
Protocols for Loading of Peptide Synthesis Resins

Analysis Note

Color (visual): white to light yellow to beige


Appearance of substance (visual): beadsLoading (determined from the substitution of the Fmoc-Leu loaded resin): 1.20 - 1.50 mmol/g


Swelling Volume (in DMF): lot specific result


Swelling Volume (in CH₂Cl₂): lot specific result


Total swelling volume acc. Houben Weyl (in CH2Cl2): ≥ 5.2 ml/g


Total swelling volume acc. Houben Weyl (in DMF): ≥ 5.2 ml/g


The polyer matrix is copoly (styrene-1 % DVB), 100 - 200 mesh

Other Notes

Replaces: 01-64-0014

Legal Information

Novabiochem is a registered trademark of Merck KGaA, Darmstadt, Germany


Storage Class

11 - Combustible Solids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Global Trade Item Number

SKUGTIN
855002000004027269109134
855002100004027269690618
855002002504027269109158
855002500004027269689087
855002010004027269109165
855002000504027269109141