Skip to Content

8.53047

Boc-Ile-OH . 0.5 H2O

Novabiochem®

Synonym(s):

Boc-Ile-OH . 0.5 H₂O, N-α-t.-Boc-L-isoleucine hemihydrate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C11H21NO4 · 0.5H2O
CAS Number:
Molecular Weight:
240.30
MDL number:
UNSPSC Code:
12352209
NACRES:
NA.22
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

product line

Novabiochem®

assay

≥98% (TLC)

form

powder

reaction suitability

reaction type: Boc solid-phase peptide synthesis

manufacturer/tradename

Novabiochem®

application(s)

peptide synthesis

storage temp.

2-30°C

SMILES string

N([C@@H]([C@H](CC)C)C(=O)O)C(=O)OC(C)(C)C.O

InChI

1S/C11H21NO4.H2O/c1-6-7(2)8(9(13)14)12-10(15)16-11(3,4)5;/h7-8H,6H2,1-5H3,(H,12,15)(H,13,14);1H2/t7-,8-;/m0./s1

InChI key

YLYHCKPXZULAEG-WSZWBAFRSA-N

General description

Standard building block for introduction of isoleucine amino-acid residues by Boc SPPS.

Associated Protocols and Technical Articles
Cleavage and Deprotection Protocols for Boc SPPS

Analysis Note

Color (visual): white to slight yellow to beige
Appearance of substance (visual): powder
Identity (IR): passes test
Optical rotation α 25/D (c=2 AcOH): +1.0 - +3.0 °
Purity (TLC(011A)): ≥ 98 %
Purity (TLC(0811)): ≥ 98 %
Solubility (1 mmole in 2 ml CH₂Cl₂): clear soluble

To see the solvent systems used for TLC of Novabiochem® products please click here.

Other Notes

Replaces: 04-12-0032

Legal Information

Novabiochem is a registered trademark of Merck KGaA, Darmstadt, Germany


Storage Class

11 - Combustible Solids

wgk

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library