Skip to Content
Merck

8.52374

Fmoc-D-Ile-OH

Novabiochem®

Synonym(s):

Fmoc-D-Ile-OH, N-α-Fmoc-D-Isoleucine

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C21H23NO4
CAS Number:
Molecular Weight:
353.41
UNSPSC Code:
12352209
NACRES:
NA.22
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

product line

Novabiochem®

form

powder

reaction suitability

reaction type: Fmoc solid-phase peptide synthesis

manufacturer/tradename

Novabiochem®

application(s)

peptide synthesis

functional group

Fmoc

storage temp.

2-30°C

SMILES string

N([C@H]([C@@H](CC)C)C(=O)O)C(=O)OCC1c2c(cccc2)c3c1cccc3

InChI

1S/C21H23NO4/c1-3-13(2)19(20(23)24)22-21(25)26-12-18-16-10-6-4-8-14(16)15-9-5-7-11-17(15)18/h4-11,13,18-19H,3,12H2,1-2H3,(H,22,25)(H,23,24)/t13-,19-/m1/s1

InChI key

QXVFEIPAZSXRGM-BFUOFWGJSA-N

General description

Standard building block for introduction of D-isoleucine by Fmoc SPPS.

Associated Protocols and Technical Articles
Cleavage and Deprotection Protocols for Fmoc SPPS

Analysis Note

Colour (visual): white to slight yellow to beige
Appearance of substance (visual): powder
Identity (IR): passes test
Enantiomeric purity: ≥ 98.0 % (a/a)
Purity (TLC(CMA2)): ≥ 98 %
Assay (HPLC, area%): ≥ 98.0 % (a/a)
Solubility (1 mmole in 2 ml DMF): clear soluble

To see the solvent systems used for TLC of Novabiochem® products please click here.

Legal Information

Novabiochem is a registered trademark of Merck KGaA, Darmstadt, Germany


Still not finding the right product?

Explore all of our products under Fmoc-D-Ile-OH


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library