Skip to Content
Merck

8.52347

Fmoc-Tyr(SO3nP)-OH

≥95.0% (HPLC), for peptide synthesis, Novabiochem®

Synonym(s):

Fmoc-Tyr(SO3nP)-OH, N-Fmoc-O-(2,2 dimethylpropylsulfo)-L-tyrosine

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C29H31NO8S
CAS Number:
Molecular Weight:
553.62
MDL number:
NACRES:
NA.22
UNSPSC Code:
12352209
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

Fmoc-Tyr(SO3nP)-OH, Novabiochem®

Quality Segment

product line

Novabiochem®

assay

≥95.0% (HPLC)

form

powder

reaction suitability

reaction type: Fmoc solid-phase peptide synthesis

manufacturer/tradename

Novabiochem®

application(s)

peptide synthesis

functional group

sulfonic acid

storage temp.

-10 to -25°C

General description

A novel derivative for the synthesis of sulfotyrosine-containing peptides by Fmoc SPPS. The sulfate neopentyl group is stable to TFA, and thus protects the sulfotyrosine residue from degradation during the cleavage reaction, but is easily cleaved post-cleavage by treatment of the peptide with aqueous ammonium acetate [1].

Associated Protocols and Technical Articles
Cleavage and Deprotection Protocols for Fmoc SPPS
Guide to Selection of Orthogonally -Protected Building Blocks for Fmoc SPPS

Literature references

[1] L. S. Simpson & T. S. Widlanski (2006) J. Am. Chem. Soc., 128, 1605.
[2] L. S. Simpson, et al. (2009) Chemistry & Biology, 16, 153.

Analysis Note

Color (visual): white to slight yellow to beige
Appearance of substance (visual): powder
Identity (IR): passes test
Enantiomeric purity: ≥ 99.0 % (a/a)
Purity (DC(BAW0)): ≥ 97 %
Assay (HPLC, area%): ≥ 95.0 % (a/a)
Solubility (1 mmole in 4 ml DMF): clearly soluble

To see the solvent systems used for TLC of Novabiochem® products please click here.

Legal Information

Novabiochem is a registered trademark of Merck KGaA, Darmstadt, Germany


Storage Class

11 - Combustible Solids

wgk

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library