Skip to Content
Merck

8.52011

Fmoc-Leu-OH

Novabiochem®

Synonym(s):

Fmoc-Leu-OH, N-α-Fmoc-L-leucine

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C21H23NO4
CAS Number:
Molecular Weight:
353.41
UNSPSC Code:
12352209
EC Index Number:
252-662-7
NACRES:
NA.22
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

product line

Novabiochem®

assay

≥98% (TLC), ≥98.0% (acidimetric), ≥99.0% (HPLC)

form

powder

reaction suitability

reaction type: Fmoc solid-phase peptide synthesis

manufacturer/tradename

Novabiochem®

mp

145-153 °C

application(s)

peptide synthesis

functional group

Fmoc

storage temp.

2-30°C

SMILES string

N(C(CC(C)C)C(=O)O)C(=O)OCC1c2c(cccc2)c3c1cccc3

InChI

1S/C21H23NO4/c1-13(2)11-19(20(23)24)22-21(25)26-12-18-16-9-5-3-7-14(16)15-8-4-6-10-17(15)18/h3-10,13,18-19H,11-12H2,1-2H3,(H,22,25)(H,23,24)

InChI key

CBPJQFCAFFNICX-UHFFFAOYSA-N

General description

High purity Fmoc-protected amino acid for research and process production of peptides, with very low levels of dipeptide, free-amino acids and acetic acid impurities. Standard building block for introduction of leucine amino-acid residues by Fmoc SPPS

Associated Protocols and Technical Articles
Fmoc-amino acids for Peptide Production
Cleavage and Deprotection Protocols for Fmoc SPPS

Application

  • Fmoc-protection efficiency: Addressing the efficiency and recrystallization needs in the Fmoc-protection process (R Yoshino et al., 2017).
  • Isotopic labeling for NMR: Development of 17O isotopic labeling of N-Formyl-MLF-OH and related building blocks, enhancing NMR spectroscopy applications (M Ha et al., 2021).
  • Perovskite solar cell modification: Investigating the role of Fmoc-Leu-OH in modifying perovskite film to enhance solar cell efficiency (J Song et al., 2022).

Analysis Note

Colour (visual): white to off white
Appearance of substance (visual): powder
Colour index (0,5 M in DMF): ≤ 150 Hazen
Identity (IR): passes test
Enantiomeric purity: ≥ 99.8 % (a/a)
Purity (HPLC): ≥ 99.0 % (a/a)
Fmoc-ß-Ala-OH (HPLC): ≤ 0.1 % (a/a)
Fmoc-ß-Ala-Leu-OH (HPLC): ≤ 0.1 % (a/a)
Fmoc-Leu-Leu-OH (HPLC): ≤ 0.1 % (a/a)
Fmoc-Ile-OH (GC): ≤ 0.1 % (a/a)
Assay free amino acid (GC): ≤ 0.2 %
Purity (TLC(011A)): ≥ 98 %
Purity (TLC(0811)): ≥ 98 %
Solubility (25 mmole in 50 ml DMF): clearly soluble
Assay (acidimetric): ≥ 98.0 %
Water (K. F.): ≤ 2.0 %
Ethyl acetate (HS-GC): ≤ 0.5 %
Acetate (IC): ≤ 0.02 %

To see the solvent systems used for TLC of Novabiochem® products please click here.

Other Notes

Replaces: 04-12-1025

Legal Information

Novabiochem is a registered trademark of Merck KGaA, Darmstadt, Germany


Still not finding the right product?

Explore all of our products under Fmoc-Leu-OH


Storage Class

11 - Combustible Solids

wgk

WGK 3



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library






Global Trade Item Number

SKUGTIN
852011050004027269179564
852011500004027269221546
852011002504027269085056
852011901504027269242497
852011010004027269085063
852011901004027269221362
852011888904027269353506
852011025004027269206963
852011102404061841205258
852011200004027269232740
852011902504027269242480
852011000004027269085032
852011100004027269179571