Skip to Content
Merck

8.52010

Fmoc-Ile-OH

Novabiochem®

Synonym(s):

Fmoc-Ile-OH, N-α-Fmoc-L-isoleucine

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C21H23NO4
CAS Number:
Molecular Weight:
353.41
UNSPSC Code:
12352209
EC Index Number:
276-255-9
NACRES:
NA.22
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

product line

Novabiochem®

assay

≥98% (TLC), ≥98.0% (acidimetric), ≥99.0% (HPLC)

form

powder

reaction suitability

reaction type: Fmoc solid-phase peptide synthesis

manufacturer/tradename

Novabiochem®

mp

143-145 °C

application(s)

peptide synthesis

functional group

Fmoc

storage temp.

2-30°C

SMILES string

N([C@@H]([C@@H](CC)C)C(=O)[O-])C(=O)OCC1c2c(cccc2)c3c1cccc3

InChI

1S/C21H23NO4/c1-3-13(2)19(20(23)24)22-21(25)26-12-18-16-10-6-4-8-14(16)15-9-5-7-11-17(15)18/h4-11,13,18-19H,3,12H2,1-2H3,(H,22,25)(H,23,24)/p-1/t13-,19+/m1/s1

InChI key

QXVFEIPAZSXRGM-YJYMSZOUSA-M

General description

High purity Fmoc-protected amino acid for research and process production of peptides, with very low levels of dipeptide, free-amino acids and acetic acid impurities.
Standard building block for introduction of isoleucine amino-acid residues by Fmoc SPPS

Associated Protocols and Technical Articles
Fmoc-amino acids for Peptide Production
Cleavage and Deprotection Protocols for Fmoc SPPS

Application


  • Total synthesis of teixobactin: This article describes the challenges and strategies in synthesizing teixobactin, a promising new antibiotic, highlighting the role of Fmoc-Ile-OH in the synthesis process (Jin et al., 2016).

  • Synthesis and structure–activity relationship studies of teixobactin analogues: Discusses the synthesis of teixobactin analogues using Fmoc-Ile-OH, providing insights into developing more effective antibiotics (Wu et al., 2017).


Analysis Note

Color (visual): white to off white
Appearance of substance (visual): powder
Color index (0,5 M in DMF): ≤ 150 Hazen
Identity (IR): passes test
Enantiomeric purity: ≥ 99.7 % (a/a)
Purity (HPLC): ≥ 99.0 % (a/a)
Fmoc-ß-Ala-OH (HPLC): ≤ 0.1 % (a/a)
Fmoc-ß-Ala-Ile-OH (HPLC): ≤ 0.1 % (a/a)
Fmoc-Ile-Ile-OH (HPLC): ≤ 0.1 % (a/a)
Fmoc-Leu-OH (GC): ≤ 0.1 % (a/a)
Assay free amino acid (GC): ≤ 0.2 %
Purity (TLC(011A)): ≥ 98 %
Purity (TLC(0811)): ≥ 98 %
Solubility (25 mmole in 50 ml DMF): clearly soluble
Assay (acidimetric): ≥ 98.0 %
Water (K. F.): ≤ 2.0 %
Ethyl acetate (HS-GC): ≤ 0.5 %
Acetate (IC): ≤ 0.02 %

To see the solvent systems used for TLC of Novabiochem® products please click here.

Other Notes

Replaces: 04-12-1024

Legal Information

Novabiochem is a registered trademark of Merck KGaA, Darmstadt, Germany


Still not finding the right product?

Explore all of our products under Fmoc-Ile-OH


Storage Class

11 - Combustible Solids

wgk

WGK 3



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library






Global Trade Item Number

SKUGTIN
852010025004027269206956
852010902504027269242473
852010000004027269084943
852010025404061841205241
852010500004027269221522
852010010004027269084974
852010200004027269232733
852010050004027269179229
852010901004027269221539
852010002504027269084967
852010100004027269179236
852010901504027269242466
852010888904027269352714