Skip to Content
Merck

8.52005

Fmoc-Asp(OtBu)-OH

Novabiochem®

Synonym(s):

Fmoc-Asp(OtBu)-OH, N-α-Fmoc-L-aspartic acid β-t.-butyl ester

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C23H25NO6
CAS Number:
Molecular Weight:
411.45
UNSPSC Code:
12352209
EC Index Number:
276-251-7
NACRES:
NA.22
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

product line

Novabiochem®

assay

≥98% (TLC), ≥98.0% (acidimetric), ≥99.0% (HPLC)

form

powder

reaction suitability

reaction type: Fmoc solid-phase peptide synthesis

manufacturer/tradename

Novabiochem®

mp

143-150 °C

application(s)

peptide synthesis

functional group

carboxylic acid

storage temp.

2-30°C

SMILES string

N([C@@H](CC(=O)OC(C)(C)C)C(=O)[O-])C(=O)OCC1c2c(cccc2)c3c1cccc3

InChI

1S/C23H25NO6/c1-23(2,3)30-20(25)12-19(21(26)27)24-22(28)29-13-18-16-10-6-4-8-14(16)15-9-5-7-11-17(15)18/h4-11,18-19H,12-13H2,1-3H3,(H,24,28)(H,26,27)/p-1/t19-/m0/s1

InChI key

FODJWPHPWBKDON-IBGZPJMESA-M

General description

High purity Fmoc-protected amino acid for research and process production of peptides, with very low levels of dipeptide, free-amino acids and acetic acid impurities.
Standard building block for introduction of aspartic acid amino-acid residues by Fmoc SPPS

Associated Protocols and Technical Articles
Cleavage and Deprotection Protocols for Fmoc SPPS

Application


  • Silybin liver research: Silybin A and B, as part of the silymarin complex, are extensively studied for their hepatoprotective properties. They play a crucial role in liver health by mitigating oxidative stress and enhancing regenerative processes in hepatic tissue (Cai et al., 2024).

  • Silybin pharmacological research: The pharmacological effects of silybin, particularly its roles in inhibiting inflammation and modulating drug metabolizing enzymes, make it a valuable compound in therapeutic research, particularly for its benefits in hepatology and oncology (Hammad et al., 2024).


Analysis Note

Colour (visual): white to off white
Appearance of substance (visual): powder
Colour index (0,5 M in DMF): ≤ 150 Hazen
Identity (IR): passes test
Enantiomeric purity: ≥ 99.8 % (a/a)
Purity (HPLC): ≥ 99.0 % (a/a)
Fmoc-ß-Ala-OH (HPLC): ≤ 0.1 % (a/a)
Fmoc-ß-Ala-Asp (OtBu)-OH (HPLC): ≤ 0.1 % (a/a)
Fmoc-Asp(OtBu)-Asp(OtBu)-OH (HPLC): ≤ 0.1 % (a/a)
Fmoc-Asp-OH (HPLC): ≤ 0.1 % (a/a)
Assay free amino acid (GC): ≤ 0.2 %
Purity (TLC(011A)): ≥ 98 %
Purity (TLC(0811)): ≥ 98 %
Solubility (25 mmole in 50 ml DMF): clearly soluble
Assay (acidimetric): ≥ 98.0 %
Water (K. F.): ≤ 2.0 %
Ethyl acetate (HS-GC): ≤ 0.5 %
Acetate (IC): ≤ 0.02 %

To see the solvent systems used for TLC of Novabiochem® products please click here.

Other Notes

Replaces: 04-12-1013

Legal Information

Novabiochem is a registered trademark of Merck KGaA, Darmstadt, Germany


Still not finding the right product?

Explore all of our products under Fmoc-Asp(OtBu)-OH


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library






Global Trade Item Number

SKUGTIN
852005025404061841364764
852005002504027269084554
852005100004027269179175
852005010004027269084561
852005902504027269242190
852005000004027269084530
852005050004027269179168
852005888904027269352110
852005025004027269206925
852005500004027269221461
852005901504027269242176
852005200004027269232665
852005901004027269221478