Skip to Content
Merck

8.22342

Trichloroacetic acid

for synthesis

Synonym(s):

Trichloroacetic acid

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
CCl3CO2H
CAS Number:
Molecular Weight:
163.39
UNSPSC Code:
12352106
EC Index Number:
200-927-2
NACRES:
NA.22
MDL number:
Assay:
≥98% (acidimetric)
Form:
solid
Grade:
synthesis grade
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

synthesis grade

Quality Segment

vapor pressure

1 hPa ( 20 °C)

assay

≥98% (acidimetric)

form

solid

autoignition temp.

711 °C

potency

3320 mg/kg LD50, oral (Rat)

pH

<1 (20 °C, 50 g/L in H2O)

mp

54-56 °C

transition temp

flash point >110 °C

solubility

1300 g/L

density

1.63 g/cm3 at 20 °C

bulk density

900 kg/m3

storage temp.

2-30°C

SMILES string

ClC(Cl)(Cl)C(=O)[O-].[H+]

InChI

1S/C2HCl3O2/c3-2(4,5)1(6)7/h(H,6,7)

InChI key

YNJBWRMUSHSURL-UHFFFAOYSA-N

Application

  • Stable dechlorination of Trichloroacetic Acid (TCAA) to acetic acid catalyzed by palladium nanoparticles: This research discusses a method for dechlorinating TCAA to acetic acid, crucial during the COVID-19 pandemic for disinfection byproducts (Cai et al., 2021).

Analysis Note

Assay (acidimetric): ≥ 98.0 %
Identity (IR): passes test


Still not finding the right product?

Explore all of our products under Trichloroacetic acid


Pictograms

CorrosionEnvironment

Signal Word

Danger

Hazard Codes

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Skin Corr. 1A

WGK

WGK 2

Flash Point (°F)

>235.4 °F - closed cup

Flash Point (°C)

> 113 °C - closed cup

Storage Class

8A - Combustible corrosive hazardous materials



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library