Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
NACRES:
NA.22
UNSPSC Code:
12352107
Assay:
19.0 - 21.0% %
Form:
liquid
Quality Segment
assay
19.0 - 21.0% %
form
liquid
reaction suitability
core: ammonium
pH
>13 (20 °C in H2O)
bp
102 °C/1013 hPa
mp
<-25 °C
transition temp
flash point >110 °C
density
1.01 g/cm3 at 20 °C
storage temp.
2-30°C
Application
- Mizoroki-Heck coupling: It serves as a solid, easy-to-handle ethylene precursor in Mizoroki-Heck coupling reactions, highlighting its application in facilitating complex organic synthesis processes (Papaplioura et al., 2024).
- Pharmacokinetic parameter prediction: It is involved in the synthesis and characterization of tetraalkylammonium salts for the pharmaceutical application of non-steroidal anti-inflammatory drugs, assisting in the in silico prediction of their pharmacokinetic parameters (Rybczyńska and Sikorski, 2023).
Analysis Note
Assay (acidimetric): 19.0 - 21.0 %
Density (d 20 °C/ 4 °C): 1.006 - 1.011
Density (d 20 °C/ 4 °C): 1.006 - 1.011
signalword
Danger
hcodes
Hazard Classifications
Acute Tox. 4 Dermal - Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1 - STOT RE 1 Dermal - STOT SE 1
target_organs
Central nervous system, thymus,Liver
Storage Class
6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects
flash_point_f
>230.0 °F
flash_point_c
> 110 °C
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.


