Skip to Content
Merck

8.20993

Phenylacetic acid

for synthesis

Synonym(s):

Phenylacetic acid

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C8H8O2
CAS Number:
Molecular Weight:
136.15
UNSPSC Code:
12352106
EC Index Number:
203-148-6
NACRES:
NA.22
MDL number:
Assay:
≥99.0% (HPLC)
Form:
solid
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


vapor pressure

1.33 hPa ( 97 °C)

Quality Segment

assay

≥99.0% (HPLC)

form

solid

potency

>5000 mg/kg LD50, oral (Rat), >5000 mg/kg LD50, skin (Rabbit)

bp

265 °C/1013 hPa

mp

74-77 °C

transition temp

flash point 132 °C

solubility

1.58 g/L

density

1.091 g/cm3 at 97 °C

storage temp.

2-30°C

SMILES string

OC(=O)Cc1ccccc1

InChI

1S/C8H8O2/c9-8(10)6-7-4-2-1-3-5-7/h1-5H,6H2,(H,9,10)

InChI key

WLJVXDMOQOGPHL-UHFFFAOYSA-N

Analysis Note

Assay (HPLC, area%): ≥ 99.0 % (a/a)
Melting range (lower value): ≥ 74 °C
Melting range (upper value): ≤ 77 °C
Identity (IR): passes test


Still not finding the right product?

Explore all of our products under Phenylacetic acid


pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Eye Irrit. 2

Storage Class

11 - Combustible Solids

wgk

WGK 1

flash_point_f

269.6 °F

flash_point_c

132 °C



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library


Related Content

PowerPoint Template





Global Trade Item Number

SKUGTIN
820993025004022536471266
820993100004022536471273
820993000004022536471242