8.20918
2,5-Norbornadiene
(stabilised) for synthesis
Synonym(s):
2,5-Norbornadiene, Bicyclo[2.2.1]hepta-2,5-diene
About This Item
vapor pressure
69 hPa ( 20 °C)
Quality Level
Assay
≥97.0% (GC)
form
liquid
autoignition temp.
350 °C
expl. lim.
1.6-6.3 % (v/v)
mp
-20 °C
transition temp
flash point -11 °C
density
0.906 g/cm3 at 20 °C
storage temp.
2-30°C
SMILES string
C21CC(C=C2)C=C1
InChI
1S/C7H8/c1-2-7-4-3-6(1)5-7/h1-4,6-7H,5H2
InChI key
SJYNFBVQFBRSIB-UHFFFAOYSA-N
Application
- Radical Polymerization: Study on the radical polymerization of 2,5-norbornadienes containing ester groups, examining the unique polymerization behaviors due to their rigid multicyclic structure (M Shiotsuki et al., 2014).
- Sequential Double C–H Functionalization: Application of 2,5-norbornadiene in flow reactors for efficient sequential double C–H functionalization, demonstrating advanced synthesis techniques (H Kim et al., 2018).
- Addition Copolymerization: Research on the addition copolymerization of silicon-containing tricyclononene with 2,5-norbornadiene dimer, exploring new materials for potential industrial applications (DA Alentiev et al., 2019).
- Ro-vibrational Spectroscopy: Investigation of the ro-vibrational spectrum of 2,5-norbornadiene, enhancing understanding of its molecular vibrations and potential applications in materials science (RL Sams, TA Blake, 2020).
- Hydroformylation in Biphasic Systems: Study on the hydroformylation of 2,5-norbornadiene in organic/aqueous two-phase systems, highlighting the role of cationic surfactants in enhancing reaction efficiency (X Yang et al., 2016).
Analysis Note
Density (d 20 °C/ 4 °C): 0.905 - 0.907
Identity (IR): passes test
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Flam. Liq. 2
Storage Class Code
3 - Flammable liquids
WGK
WGK 2
Flash Point(F)
12.2 °F
Flash Point(C)
-11 °C
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Articles
MOST offers controlled solar energy harvesting and storage, addressing global energy demands with improved storage techniques.
MOST offers controlled solar energy harvesting and storage, addressing global energy demands with improved storage techniques.
MOST offers controlled solar energy harvesting and storage, addressing global energy demands with improved storage techniques.
MOST offers controlled solar energy harvesting and storage, addressing global energy demands with improved storage techniques.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service