Skip to Content
Merck

820463

Probumin® Bovine Serum Albumin Diagnostic Grade

pkg of 1 L, 29-31 g/dL protein (Biuret), USA origin, Tested for protease, enzyme immunoassay: suitable, ELISA: suitable, Serology: suitable

Synonym(s):

Albumin solution from bovine serum, BSA solution, Albumin, BSA, Bovine Serum, Bovine Serum Albumin, Fraction V

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

CAS Number:
MDL number:
UNSPSC Code:
41141801
NACRES:
NA.28
Form:
solution (30%)
Assay:
29-31 g/dL protein basis (biuret)
Mol wt:
~66 kDa
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

Probumin® Bovine Serum Albumin Diagnostic Grade, solution (30%), pkg of 1 L

Quality Segment

assay

29-31 g/dL protein basis (biuret)

form

solution (30%)

shelf life

48 mo.

mol wt

~66 kDa

purified by

heat shock fractionation

packaging

pkg of 1 L

manufacturer/tradename

Probumin®

origin

USA origin

concentration

29-31 g/dL protein (biuret)

technique(s)

ELISA: suitable, enzyme immunoassay: suitable

dilution

(Serology)

impurities

Tested for IgG

pH

7.2-7.4

foreign activity

Bluetongue, none detected, Tested for Protease , Tested for protease , Vesicular Stomatitis Virus, none detected, microbial content by USP tested , preservative, free

storage temp.

2-8°C

General description

Albumin is one of the most extensively utilized proteins in the medical industry. Its availability, inherent stability, and ligand binding capability make albumin a critical raw material in the development and production of many diagnostic tests. With over 65 years of protein fractionation expertise, we are an experienced manufacturer of consistent bovine serum albumin products for the diagnostic and cell culture markets. Our proprietary heat shock and cold solvent fractionation processing technology allows for the delivery of products with high-quality, performance, and reliability.

Application

Probumin® BSA products are fit for use as blockers, protein/antibody (conjugate) stabilizers, and in immunoassays (EIA, RIA and serology systems).

Features and Benefits

  • High-quality and reliability. Assurance of consistency in performance and supply.
  • Proprietary heat shock and cold solvent fractionation process optimizes the protein yield, purity, and efficacy of BSA. Large batch capacity.
  • Safe serum sourcing from USDA-licensed facilities exclusively from U.S. cattle.
  • Processed to eliminate or reduce proteases.

Legal Information

Probumin is a registered trademark of Merck KGaA, Darmstadt, Germany


Still not finding the right product?


Storage Class

10 - Combustible liquids

wgk

WGK 1



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library