Skip to Content
Merck

8.20265

2-Chloroethylammonium chloride

for synthesis

Synonym(s):

2-Chloroethylammonium chloride, 2-Chloroethylamine hydrochloride, β-Aminoethyl chloride hydrochloride

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C2H6ClN · xHCl
CAS Number:
Molecular Weight:
79.53 (free base basis)
UNSPSC Code:
12352107
EC Index Number:
212-793-2
NACRES:
NA.22
MDL number:
Form:
crystals
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

form

crystals

potency

>2000 mg/kg LD50, oral (Rat)

pH

2-3 (20 °C, 5666 g/L in H2O)

mp

143-146 °C

solubility

5666 g/L

bulk density

750‑850 kg/m3

storage temp.

2-30°C

SMILES string

[Cl-].ClCC[N+H3]

InChI

1S/C2H6ClN.ClH/c3-1-2-4;/h1-2,4H2;1H

InChI key

ONRREFWJTRBDRA-UHFFFAOYSA-N

Analysis Note

Assay (argentometric): ≥ 99.0 %
Identity (IR): passes test


pictograms

CorrosionHealth hazard

signalword

Danger

Hazard Classifications

Eye Dam. 1 - Met. Corr. 1 - Muta. 2 - Skin Corr. 1B

Storage Class

8A - Combustible, corrosive hazardous materials

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library