Skip to Content

8.20150

Dimethyl succinate

for synthesis

Synonym(s):

Dimethyl succinate, Succinic acid dimethyl ester

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C6H10O4
CAS Number:
Molecular Weight:
146.14
UNSPSC Code:
12352108
EC Index Number:
203-419-9
NACRES:
NA.22
MDL number:
Assay:
≥95.0% (GC)
Form:
liquid
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


vapor pressure

0.2 hPa ( 20 °C)

Quality Segment

assay

≥95.0% (GC)

form

liquid

autoignition temp.

365 °C

potency

>5000 mg/kg LD50, oral (Rat), >5000 mg/kg LD50, skin (Rabbit)

expl. lim.

1.0-8.5 % (v/v)

bp

196 °C/1013 hPa

mp

18-19 °C

transition temp

flash point 94 °C

solubility

75 g/L

density

1.12 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

O(C)C(=O)CCC(=O)OC

InChI

1S/C6H10O4/c1-9-5(7)3-4-6(8)10-2/h3-4H2,1-2H3

InChI key

MUXOBHXGJLMRAB-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 95.0 % (a/a)
Density (d 20 °C/ 4 °C): 1.119 - 1.120
Identity (IR): passes test
Due to its specific melting range the product may be solid, liquid, a solidified melt or a supercooled melt.


Still not finding the right product?

Explore all of our products under Dimethyl succinate


pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Eye Irrit. 2

Storage Class

10 - Combustible liquids

wgk

WGK 1

flash_point_f

201.2 °F - closed cup

flash_point_c

94 °C - closed cup



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library